Skip to Content
Merck
All Photos(1)

Key Documents

437891

Sigma-Aldrich

Dimethyl chlorophosphate

96%

Synonym(s):

Dimethyl phosphorochloridate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)Cl
CAS Number:
Molecular Weight:
144.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

80 °C/25 mmHg (lit.)

density

1.34 g/mL at 25 °C (lit.)

SMILES string

COP(Cl)(=O)OC

InChI

1S/C2H6ClO3P/c1-5-7(3,4)6-2/h1-2H3

InChI key

NGFFLHMFSINFGB-UHFFFAOYSA-N

Application

Dimethyl chlorophosphate may be used for the preparation of (E)-3-cyanophosphoenolpyruvate, by reaction with the potassium enolate of ethyl cyanopyruvate.[1] It may be used for the synthesis of mono- and diphosphate esters of 2-arachidonyl glyceryl ether.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juha Juntunen et al.
Journal of medicinal chemistry, 46(23), 5083-5086 (2003-10-31)
The poor aqueous solubility of 2-arachidonyl glyceryl ether (noladin ether) 2 hinders both pharmacological studies and pharmaceutical development. The synthesized mono- and diphosphate esters of noladin ether (4 and 6) considerably increased the aqueous solubility of noladin ether (>40000-fold), showed
Amanda R Wilmsmeyer et al.
Langmuir : the ACS journal of surfaces and colloids, 28(30), 10962-10967 (2012-07-12)
The fundamental interactions of dimethyl methylphosphonate (DMMP) and dimethyl chlorophosphate (DMCP) on amorphous silica nanoparticles have been investigated with transmission infrared spectroscopy and temperature-programmed desorption (TPD). DMMP and DMCP both adsorb molecularly to silica through the formation of hydrogen bonds
P Wirsching et al.
Biochemistry, 24(26), 7602-7606 (1985-12-17)
(E)-3-Cyanophosphoenolpyruvate has been synthesized by reacting dimethyl chlorophosphate with the potassium enolate of ethyl cyanopyruvate. The resulting trialkyl ester was deesterified with bromotrimethylsilane followed by potassium hydroxide. Subsequent treatment with Dowex-50-H+ resin and cyclohexylamine afforded the tricyclohexylammonium salt; only the
Minji Kang et al.
ACS applied materials & interfaces, 11(8), 7686-7694 (2019-02-16)
Quorum sensing (QS) inhibitor-based therapy is an attractive strategy to inhibit bacterial biofilm formation without excessive induction of antibiotic resistance. Thus, we designed Ca2+-binding poly(lactide- co-glycolide) (PLGA) microparticles that can maintain a sufficient concentration of QS inhibitors around hydroxyapatite (HA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service