Skip to Content
Merck
All Photos(1)

Documents

377953

Sigma-Aldrich

2-Chloro-1,3,2-dioxaphospholane 2-oxide

Synonym(s):

2-Chloro-2-oxo-1,3,2-dioxaphospholane, Ethylene glycol chlorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4ClO3P
CAS Number:
Molecular Weight:
142.48
Beilstein:
606582
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

impurities

<10% 2-Chloro-1,3,2-dioxaphospholane

refractive index

n20/D 1.45 (lit.)

bp

89-91 °C/0.8 mmHg (lit.)

mp

12-14 °C (neat) (lit.)

density

1.55 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

ClP1(=O)OCCO1

InChI

1S/C2H4ClO3P/c3-7(4)5-1-2-6-7/h1-2H2

InChI key

SBMUNILHNJLMBF-UHFFFAOYSA-N

General description

2-Chloro-1,3,2-dioxaphospholane 2-oxide (COP) is a cyclic chlorophosphate reagent that can be prepared from 2-chloro-1,3,2-dioxaphospholane by reacting with molecular oxygen .

2-Chloro-1,3,2-dioxaphospholane 2-oxide is used in esterification reactions for cyclic phosphate synthesis, also reacts with phenyl grignard reagents.

Application

2-Chloro-1,3,2-dioxaphospholane 2-oxide may be used in the synthesis of:
  • 2-methacryloyloxyethylphosphorylcholine
  • miltefosine (hexadecylphosphocholine, MT) analogs
  • phosphoric acid 2-trimethylamino-ethyl ester undec-10-enyl ester
  • uridine nucleolipid, (2′,3′-O-16-hentriacontanyliden-uridine-5′-phosphocholine, PUPC)
  • adenosine nucleoamphiphile, (2′,3′-O-16-hentriacontanyliden-adenosine-5′-phosphocholine, PAPC)
  • structurally related phospholipids which are either conformationally restricted or flexible
  • phosphatidylcholines
Reactant for:
  • Synthesis of amino-functionalized hybrid hydrocarbon/fluorocarbon double-chain phospholipid
  • Synthesis of UV-polymerizable lipids via Chabrier reaction
  • Syntheses of block copolymers of poly(aliphatic ester) with clickable polyphosphoester
  • Imprinting molecular recognition sites on multiwalled carbon nanotubes surface for electrochemical detection of insulin in real samples
  • Synthesis of a zwitterionic silane
  • Synthesis of a core-shell-corona micelle stabilized by reversible cross-linkage for intracellular drug delivery

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and tensioactive properties of PEO-b-polyphosphate copolymers
Stephanie V et al.
Royal Society of Chemistry Advances, 5, 27330-27337 (2015)
Toward the Efficient Synthesis of New Phosphopantothenate Derivatives by Using Chlorophosphate Reagents.
Pahor J, et al.
Synthesis, 48(21), 3763-3772 (2016)
Journal of the American Chemical Society, 115, 6600-6600 (1993)
Journal of the Chemical Society. Perkin Transactions 1, 2521-2521 (1993)
Phospholipid based polymer as drug release coating for cardiovascular device.
Xu J-P, et al.
Eur. Polymer J., 40(2), 291-298 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service