Skip to Content
Merck
All Photos(3)

Documents

433063

Sigma-Aldrich

Sodium methanesulfinate

technical grade, 85%

Synonym(s):

Methanesulfinic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2Na
CAS Number:
Molecular Weight:
102.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

mp

222-226 °C (dec.) (lit.)

SMILES string

[Na+].CS([O-])=O

InChI

1S/CH4O2S.Na/c1-4(2)3;/h1H3,(H,2,3);/q;+1/p-1

InChI key

LYPGDCWPTHTUDO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium methanesulfinate is an aliphatic sodium sulfinate. Conjugate addition of sodium methanesulfinate to vinyl heterocycles has been described. Cross-coupling reaction between aryl boronic acid and sodium methanesulfinate has been studied. Its stock solution was prepared from methanesulfonic acid by adding one equivalent of sodium hydroxide and diluting it to 4M.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C F Babbs et al.
Free radical biology & medicine, 6(5), 493-503 (1989-01-01)
A major impediment to the confirmation of free radical mechanisms in pathogenesis is a lack of direct, chemical evidence that oxygen centered free radicals actually arise in living tissues in quantities sufficient to cause serious damage. This investigation was conducted
Conjugate addition of sodium methanesulfinate to vinyl pyridines and diazines for the synthesis of aliphatic sulfones.
Schaaf GM, et al.
Tetrahedron Letters, 50(17), 1928-1933 (2009)
Jerry J Wu et al.
Journal of hazardous materials, 149(1), 218-225 (2007-05-01)
The present study investigates the oxidation of dimethyl sulfoxide (DMSO) by conventional ozonation and the advanced oxidation processes (AOPs). The major degradation products identified were methanesulfinate, methanesulfonate, formaldehyde, and formic acid in ozonation process. The subsequent degradation of intermediates shows
Cecilia Arsene et al.
Environmental science & technology, 36(23), 5155-5163 (2003-01-14)
Dimethyl sulfoxide (CH3S(O)CH3: DMSO) is an important product of dimethyl sulfide (CH3SCH3: DMS) photooxidation. The mechanism of the OH-radical initiated oxidation of DMSO is still highly uncertain and a major aim of recent studies has been to establish if methane
One-pot synthesis of aryl sulfoxides and sulfonium salts from sulfinic acid as a novel sulfurizing agent.
Yamamoto K, et al.
Chemical Communications (Cambridge, England), 17, 2099-2100 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service