Skip to Content
Merck
All Photos(3)

Key Documents

318183

Sigma-Aldrich

Methyl sulfate sodium salt

Synonym(s):

Monomethyl ester sulfuric acid sodium salt, Sodium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OSO3Na
CAS Number:
Molecular Weight:
134.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

impurities

<3% methanol
<5% water

mp

210 °C (lit.)

SMILES string

[Na+].COS([O-])(=O)=O

InChI

1S/CH4O4S.Na/c1-5-6(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

DZXBHDRHRFLQCJ-UHFFFAOYSA-M

Application

Methyl sulfate sodium salt can be used to synthesize:
  • Methylsulfate anion based 1,3,4-trialkyl-1,2,3-triazolium ionic liquids for use in Morita–Baylis–Hillman reaction.[1]
  • Anisole by reacting with phenol.[2]
  • Hydroxyanilino quinolines for use as RET kinase inhibitors.[3]

Reactant or reagent involved in:
  • Studying lamellar structure formation in hybrid nanomaterials created by miniemulsion
  • EPR studies of radical ions radiolytically generated from ionic liquids, used as a reference
  • Micellular studies specifically the rate-retarding effects on hydrolysis of substituted 1-benzoyl-1,2,4-triazoles and self-assembly / microstructure of mixed micelles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of anisole from phenol and sodium methyl sulfate, a byproduct from synthesis of medicine intermediates
Guoping W, et al.
Petrochemical Technology, 45(11), 1337-1337 (2016)
Synthesis of new triazolium-based ionic liquids and their use in the Morita-Baylis-Hillman reaction
F Alioune, et al.
Tetrahedron Letters, 56(36), 5128-5131 (2015)
The discovery of substituted 4-(3-hydroxyanilino)-quinolines as potent RET kinase inhibitors
Robinett R G, et al.
Bioorganic & Medicinal Chemistry Letters, 17(21), 5886-5893 (2007)
V Chatsudthipong et al.
The Journal of pharmacology and experimental therapeutics, 288(3), 993-1001 (1999-02-23)
The transport step for p-aminohippurate (PAH) from cell to lumen across the luminal membrane of rabbit proximal tubules has not been adequately defined. To examine this process more closely, we determined the effects of possible transport inhibitors and substitutes for
Sami A Goussous et al.
Journal of colloid and interface science, 488, 365-372 (2016-11-16)
Molecular scale information about the structure of surfactants at interfaces underlies their application in consumer products. In this study the non-linear optical technique of Sum Frequency Generation (SFG) vibrational spectroscopy has been used to investigate the structure and temporal behaviour

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service