Skip to Content
Merck
All Photos(2)

Key Documents

430234

Sigma-Aldrich

Bisphenol A diglycidyl ether, brominated

mol wt 350-450 g/epoxide by perchloric acid method

Synonym(s):

4,4′-(1-Methylethylidene)bis[2,6-dibromophenol] polymer with (chloromethyl)oxirane, 4,4′-(1-methylethylidene)bis(2,6-dibromophenol), Tetrabromobisphenol A-epichlorohydrin polymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

Quality Level

mol wt

350-450 g/epoxide by perchloric acid method

extent of labeling

48 wt. % Br

viscosity

1,800 cSt(lit.)

mp

55-65 °C (lit.)

transition temp

softening point 55-65 °C (Durrans)

solubility

a variety of ketone, aromatic hydrocarbon and ether alcohol solvents: soluble

SMILES string

Brc2c(c(cc(c2)C(C)(C)c3cc(c(c(c3)Br)O)Br)Br)O.ClCC1OC1

InChI

1S/C15H12Br4O2.C3H5ClO/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8;4-1-3-2-5-3/h3-6,20-21H,1-2H3;3H,1-2H2

InChI key

DEGGCMXJUHYWMD-UHFFFAOYSA-N

General description

Bisphenol A diglycidyl ether, brominated is a brominated epoxy resin that is used as a flame retardant material. It is majorly utilized as a combustible epoxy resin in the production of electronic printed boards.[1][2]

Application

Imparts flame retardancy to molding, laminating and casting systems.

Physical properties

A non-sintering material, high bromine content, low softening point.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Stockert et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 52(5-6), 408-411 (1997-05-01)
Treatment of chicken blood smears and semithin sections from Epon-embedded mouse tissues with aqueous solutions of the 3-aminonaphthalimides FA-142, FA-2043, and FA-2143 induced a strong green-yellow fluorescence of chromatin under violet or violet-blue excitation. Chromatin emission was abolished by previous
Thermal decomposition of fire retardant brominated epoxy resins cured with different nitrogen containing hardeners
Luda MP, et al.
Polymer Degradation and Stability, 92(6), 1088-1100 (2007)
A G Pogorelov et al.
Bulletin of experimental biology and medicine, 141(4), 513-519 (2006-12-13)
Electron probe microanalysis is a method for evaluation of concentrations of elements at the subcellular level, effective in studies of an individual cell in culture or suspension. Intracellular concentrations of cytoplasmic ions (K+, Na+, Cl-) most rapidly and markedly reacting
G M Kim et al.
Journal of nanoscience and nanotechnology, 2(1), 55-59 (2003-08-12)
A means of accurate control of the curvature radius of molds that are used in nanostructure replication techniques is presented. The local non-uniform growth of SiO2 at regions with high curvature is used to fabricate molds with a curvature radius
E Kirschning et al.
Journal of neuroscience research, 53(4), 465-474 (1998-08-26)
Galactocerebroside (GalC) and sulfatide are major constituent lipids in vertebrate myelin. Their precise immunolocalization in electron microscopy so far has been hampered by the fact that lipids are not immobilized by chemical fixation and thus get extracted during dehydration with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service