Skip to Content
Merck
All Photos(1)

Key Documents

15138

Supelco

Bisphenol A diglycidyl ether

analytical standard

Synonym(s):

2,2-Bis[4-(glycidyloxy)phenyl]propane, 4,4′-Isopropylidenediphenol diglycidyl ether, BADGE, D.E.R. 332

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O4
CAS Number:
Molecular Weight:
340.41
Beilstein:
299026
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

40-47 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(c1ccc(OCC2CO2)cc1)c3ccc(OCC4CO4)cc3

InChI

1S/C21H24O4/c1-21(2,15-3-7-17(8-4-15)22-11-19-13-24-19)16-5-9-18(10-6-16)23-12-20-14-25-20/h3-10,19-20H,11-14H2,1-2H3

InChI key

LCFVJGUPQDGYKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 195.6 Å2 [M+H]+ and 174.5 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 15138 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.
Standard for determining toxic monomers released from polymers of the inner coating of cans.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

507.2 - 514.4 °F - closed cup

Flash Point(C)

264 - 268 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandros G Asimakopoulos et al.
Journal of chromatography. A, 1324, 141-148 (2013-12-10)
A liquid-liquid extraction (LLE; ethyl acetate) protocol, followed by liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) methodology, was developed for the determination of 19 compounds, including bisphenol A diglycidyl ethers (BADGEs; industrial ethers), benzophenone-type UV filters (BP-UV filters; precursors and
Chromatographia, 34, 67-67 (1992)
Mona F Mahmoud et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 51, 114-122 (2012-09-20)
Cisplatin-induced nephrotoxicity is a serious problem that limits its use in cancer treatment. The present study aimed to investigate the renal protective capacity of pioglitazone to reduce the cisplatin- induced nephrotoxicity. The underlying suggested mechanism(s) and whether this nephroprotective effect
J Míguez et al.
Food chemistry, 135(3), 1310-1315 (2012-09-08)
A new and simple liquid chromatography tandem mass-spectrometry method for the determination of different bisphenol A (BPA) derivatives such as bisphenol A diglycidyl ether (BADGE), bisphenol F diglycidyl ether (BFDGE) and their reaction products with water and hydrochloric acid in
Natchanun Leepipatpiboon et al.
Journal of chromatography. A, 1073(1-2), 331-339 (2005-05-25)
A gradient reversed-phase liquid chromatography with fluorescence detection method for simultaneous identification and quantification of bisphenol-A-diglycidyl ether (BADGE), bisphenol-F-diglycidyl ether (BFDGE), and their 10 derivatives in food matrixes was developed and validated for the analysis of oil-in-water- and aqueous-based foodstuffs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service