Skip to Content
Merck
All Photos(2)

Key Documents

427519

Sigma-Aldrich

2-Methylglutaronitrile

99%

Synonym(s):

α-Methylglutarodinitrile, α-Methylglutaronitrile, 1,3-Dicyanobutane, 2,4-Dicyanobutane, 2-Methylpentanedinitrile, Diacrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH(CH3)CH2CH2CN
CAS Number:
Molecular Weight:
108.14
Beilstein:
1741955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

impurities

<0.1% benzene

refractive index

n20/D 1.434 (lit.)

bp

269-271 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble

density

0.95 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CC(CCC#N)C#N

InChI

1S/C6H8N2/c1-6(5-8)3-2-4-7/h6H,2-3H2,1H3

InChI key

FPPLREPCQJZDAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylglutaronitrile (MGN) is an aliphatic dinitrile. It is a high boiling solvent mainly used in the fiber industry. It is formed as a byproduct during the synthesis of adiponitrile. Synthesis of β-picoline using a mixture of 2-methylglutaronitrile and 2-ethylsuccinonitrile in the presence of silica-supported palladium, rhodium and platinum catalysts by vapour phase hydrogenation has been studied. Its core electron binding energy (CEBE) has been calculated via density functional theory (DFT). It has been reported to undergo hydrolysis catalyzed by nitrilase to form 4-cyanopentanoic acid.

Application

2-Methylglutaronitrile (MGN) may be used in the synthesis of 1,5-dimethyl-2-piperidone (1,5-DMPD) by chemoenzymatic process.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ?-picoline from 2-methylglutaronitrile over supported noble metal catalysts II. Influence of operation parameters.
Lanini S and Prins R.
Applied Catalysis A: General, 137(2), 307-326 (1996)
Optimization of an immobilized-cell biocatalyst for production of 4-cyanopentanoic acid.
Hann EC, et al.
Organic Process Research & Development, 6(4), 492-496 (2002)
Accurate density functional calculation of core electron binding energies: Part V: application to nitriles. Model molecules for polyacrylonitrile revisited.
Bureau C, et al.
Journal of Electron Spectroscopy and Related Phenomena, 83(2), 227-234 (1997)
Chemoenzymatic production of 1, 5-dimethyl-2-piperidone.
Cooling FB, et al.
Journal of Molecular Catalysis. B, Enzymatic, 11(4), 295-306 (2001)
A nitrilase from a metagenomic library acts regioselectively on aliphatic dinitriles.
Bayer S, et al.
Applied Microbiology and Biotechnology, 89(1), 91-98 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service