Skip to Content
Merck
All Photos(1)

Key Documents

424749

Sigma-Aldrich

trans-β-Methyl-β-nitrostyrene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(CH3)NO2
CAS Number:
Molecular Weight:
163.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
99%

Quality Level

Assay

99%

form

solid

mp

63-65 °C (lit.)

functional group

amine
nitro
phenyl

storage temp.

2-8°C

SMILES string

[H]\C(=C(\C)[N+]([O-])=O)c1ccccc1

InChI

1S/C9H9NO2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-7H,1H3/b8-7+

InChI key

WGSVFWFSJDAYBM-BQYQJAHWSA-N

General description

trans-β-Methyl-β-nitrostyrene (1-phenyl-2-nitropropene), a nitrostyrene derivative is an α,β-disubstituted nitroalkene. It has been synthesized by reacting benzaldehyde with nitroethane and butylamine. Spectroscopic analysis of 1-phenyl-2-nitropropene has been done using FT-IR, FT-Raman, NMR and UV.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Spectroscopic (FT-IR, FT-Raman, UV and NMR) Investigation on 1-Phenyl-2-Nitropropene by Quantum Computational Calculations.
Xavier S and Periandy S.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy (2015)
Nitro alkene derivatives.
Hass HB, et al.
The Journal of Organic Chemistry, 15(1), 8-14 (1950)
Tundo P and Andraos J
Green Syntheses, 119-119 (2014)
Organocatalytic diastereo- and enantioselective sulfa-Michael addition to α,β-disubstituted nitroalkenes
Pei QL, et al.
Tetrahedron, 69, 5367-5373 (2013)
A Mori et al.
Chemical & pharmaceutical bulletin, 38(12), 3449-3451 (1990-12-01)
Microbial reduction of 1-phenyl-2-nitro-1-propene (3) was carried out using 57 strains of yeast, 40 strains of aerobic and facultatively anaerobic bacteria and 40 strains of strictly anaerobic bacteria. Nine strains of yeast (Candida tropicalis, etc.,) had the ability to reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service