414093
Poly(vinyl alcohol-co-ethylene)
ethylene 32 mol %
Synonym(s):
PVA-co-PE
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(CH2CH2)x[CH2CH(OH)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
pellets
Quality Level
melt index
3.8 g/10 min (210°C)
composition
ethylene, 32 mol %
hardness
100 (Rockwell M, ASTM D 785)
impurities
<0.7% polymerized vinyl acetate
transition temp
Tm 183 °C (DSC peak)
Tg 69 °C
solubility
DMSO, water/n-propanol (1:1), and phenol: soluble
density
1.19 g/mL at 25 °C
SMILES string
C=C.OC=C
InChI
1S/C2H4O.C2H4/c1-2-3;1-2/h2-3H,1H2;1-2H2
InChI key
UFRKOOWSQGXVKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Poly(vinyl alcohol-co-ethylene) (PVA-co-PE) is a hydrophilic biocompatible polymer that has a non-toxic nature. It has high tensile strength and good film forming ability that make it useful in the food packaging industry.
Application
PVA-co-PE can be used as a membrane in a variety of applications like filtration, biosensor and food sensors. It can also be used as a nanoporous membrane, which can potentially be used as a separator in energy vehicles and lithium ion batteries.
Other Notes
Maximum working temperature 240°C
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A novel hierarchically structured and highly hydrophilic poly (vinyl alcohol-co-ethylene)/poly (ethylene terephthalate) nanoporous membrane for lithium-ion battery separator
Xia M, et al.
Journal of Power Sources, 266(7), 29-35 (2014)
Noncrystalline nickel phosphide decorated poly (vinyl alcohol-co-ethylene) nanofibrous membrane for catalytic hydrogenation of p-nitrophenol
Liu K, et al.
Applied Catalysis. B, Environmental, 196(7), 223-231 (2016)
High performance filtration nanofibrous membranes based on hydrophilic poly (vinyl alcohol-co-ethylene) copolymer
Li M, et al.
Desalination, 329, 50-56 (2013)
An endovascular technique for treatment of high-risk iatrogenic aortic pseudoaneurysms with the ethylene vinyl alcohol copolymer onyx and aortic stent grafting.
K-M Eberhardt et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 184(12), 1163-1165 (2012-11-17)
Peter Adamczyk et al.
Neurosurgical focus, 32(5), E12-E12 (2012-04-28)
Endovascular embolization with Onyx has been increasingly used to treat intracranial and spinal dural arteriovenous fistulas (DAVFs). Several case series have been published in recent years reporting high DAVF cure rates with this technique. Although it is seldom reported, DAVF
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service