Skip to Content
Merck
All Photos(3)

Key Documents

189480

Sigma-Aldrich

Poly(vinyl acetate)

average Mw ~100,000 by GPC, beads

Synonym(s):

Polyvinyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€62.20
500 G
€239.00
1 KG
€369.00

€62.20


In StockDetails


To order products, please contact your local dealer.

Select a Size

Change View
25 G
€62.20
500 G
€239.00
1 KG
€369.00

About This Item

Linear Formula:
[CH2CH(O2CCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€62.20


In StockDetails


To order products, please contact your local dealer.

form

beads

Quality Level

mol wt

average Mw ~100,000 by GPC

viscosity

17-21 sec.(25 °C, Ford, no. 4)(lit.)

density

1.18 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl acetate) (PVAc) is a non-crystalline, atactic polymer. It is prepared from vinylacetate monomer via free radical polymerization. It is commonly usedin the food industry due to its low toxicity and excellent flexibility. PVAc films are used as emission sensors forvehicles and in oral drug delivery products.

Application

Poly(vinyl acetate) can be used as a:
  • Starting material to prepare polymer fibers for gas and humidity sensor applications. Citric acid-doped PVAc nanofibrous mats can be used to develop sensors for trimethylamine gas at room temperature.
  • Polymer coating for floating tablets showing a controlled drug delivery. PVAc is a suitable excipient to ensure reliable and safe drug release. PVAc-based matrix tablet flow due to intrinsic low density and high elasticity of polyvinyl acetate coating.
It can also be used to prepare cellulose nanofibrils(CNF)-PVAc nanocomposite adhesives by simple mixing of CNFs suspension with PVAc and polyvinyl alcohol solution. The addition of CNFs enhances the mechanical properties of the nanocomposite.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ons Chaabouni et al.
Carbohydrate polymers, 156, 64-70 (2016-11-16)
Cellulose nanofibrils (CNFs) are nanoscale cellulose produced from renewable resources with strong reinforcing potential when included in a polymer matrix. In this work, the effect of the addition of CNFs on the properties of waterborne polyvinylacetate (PVA) adhesive was investigated.
K Kolter et al.
International journal of pharmaceutics, 457(2), 470-479 (2013-10-01)
Polyvinyl acetate-based colloidal aqueous polymer dispersion Kollicoat(®) SR 30 D results in coatings characterized by moderate swelling behaviour, lipophilicity, pH-independent permeability for actives and high flexibility to withstand mechanical stress and is therefore used for controlled release coating. The colloidal
Priscileila Colerato Ferrari et al.
Carbohydrate polymers, 91(1), 244-252 (2012-10-10)
In this work pellets containing chitosan for colonic drug delivery were developed. The influence of the polysaccharide in the pellets was evaluated by swelling, drug dissolution and intestinal permeation studies. Drug-loaded pellets containing chitosan as swellable polymer were coated with
Seon Tae Kim et al.
International journal of pediatric otorhinolaryngology, 77(1), 113-116 (2012-11-08)
The purpose of this prospective study was to determine the effectiveness of polyurethane foam (PUF) and polyvinyl acetate (PA) as packing materials for reducing post-conchotomy bleeding, pain, and headaches. This study was a prospective, randomized and single-blinded controlled study. Fifty-two
Morphology of new zwitterionic copolymer matrices for sustained drug release.
Bistra Kostova et al.
Journal of controlled release : official journal of the Controlled Release Society, 148(1), e44-e45 (2011-05-03)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service