Skip to Content
Merck
All Photos(1)

Documents

413496

Sigma-Aldrich

Dodecyl vinyl ether

98%

Synonym(s):

Lauryl vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OCH=CH2
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.4382 (lit.)

bp

117-120 °C (lit.)

density

0.817 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCOC=C

InChI

1S/C14H28O/c1-3-5-6-7-8-9-10-11-12-13-14-15-4-2/h4H,2-3,5-14H2,1H3

InChI key

LAYAKLSFVAPMEL-UHFFFAOYSA-N

Application

  • Synthesis of ABA Type Block Copolymers of Poly (Ethylene Glycol) and Poly (Dodecyl Vinyl Ether): Research on cationic polymerization of dodecyl vinyl ether yielding ABA type block copolymer, used as surfactant in emulsion polymerization (YH Gursel, A Sarac, BF Senkal, 2014).
  • A sustainable polymer and coating system based on renewable raw materials: Focus on monomers like methoxybutenolide and dodecyl vinyl ether derived from biomass, used as a biobased crosslinker (JGH Hermens, T Freese, G Alachouzos, ML Lepage, 2022).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service