329800
Tri(ethylene glycol) divinyl ether
98%
Synonym(s):
3,6,9,12-Tetraoxatetradeca-1,13-diene, Triethylene glycol divinyl ether
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
H2C=CH(OCH2CH2)3OCH=CH2
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor pressure
20 mmHg ( 137 °C)
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.453 (lit.)
bp
120-126 °C/18 mmHg (lit.)
density
0.99 g/mL at 25 °C (lit.)
SMILES string
C=COCCOCCOCCOC=C
InChI
1S/C10H18O4/c1-3-11-5-7-13-9-10-14-8-6-12-4-2/h3-4H,1-2,5-10H2
InChI key
CYIGRWUIQAVBFG-UHFFFAOYSA-N
Related Categories
Application
Tri(ethylene glycol) divinyl ether is a bifunctional monomer that can be used in a variety of properties such as: formation of gel polymer electrolyte for lithium ion batteries, for the investigation of photo-initiators by cationic polymerization of vinyl ester monomers
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of novel sulfonium salts and cationic polymerization of epoxides and vinyl ether
Hyun DH, et al.
Journal of Applied Polymer Science, 103(5), 3157-3163 (2007)
Room temperature cross-linkable gel polymer electrolytes for lithium ion batteries by in situ cationic polymerization of divinyl ether
Hwang SS, et al.
Electrochemical Communications, 12(7), 916-919 (2010)
Photoinitiated cationic polymerization of vinyl ethers using substituted vinyl halides
Kahveci MU, et al.
Macromolecules, 42(13), 4443-4448 (2009)
Influence of type of zinc salts on photoinitiated living cationic polymerization of vinyl ethers
Kahveci MU, et al.
Polymer, 54(18), 4798-4801 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service