If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
408263
Pentaerythritol tetraacrylate
contains 350 ppm monomethyl ether hydroquinone as inhibitor
Synonym(s):
2,2-Bis[[(1-oxo-2-propenyl)oxy]methyl]-1,3-propanediyl ester, Tetramethylolmethane tetraacrylate
About This Item
Recommended Products
contains
350 ppm monomethyl ether hydroquinone as inhibitor
impurities
10-40% triester
refractive index
n20/D 1.487 (lit.)
density
1.19 g/mL at 25 °C (lit.)
SMILES string
C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
InChI
1S/C17H20O8/c1-5-13(18)22-9-17(10-23-14(19)6-2,11-24-15(20)7-3)12-25-16(21)8-4/h5-8H,1-4,9-12H2
InChI key
KNSXNCFKSZZHEA-UHFFFAOYSA-N
General description
Application
- To synthesize polymer electrolytes for the fabrication of lithium-sulfur batteries.
- In the formulation of UV-curable epoxy acrylate coatings, to improve their mechanical properties and performance.
- To prepare multifunctional silicone acrylate prepolymers for use in UV-curable coatings, to improve their mechanical and anti-oxygen inhibition properties.
- In the UV-curable ink formulations.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
I purchased PETA can you let me the storage temperature for this chemical
1 answer-
This product is to be stored at room temperature. Please note that this product is heat sensitive and polymerization may occur if the material is exposed to high temperatures.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service