Skip to Content
Merck
All Photos(1)

Key Documents

246794

Sigma-Aldrich

Pentaerythritol tetraacrylate, Pentaerythritol triacrylate, and Trimethylolpropane triacrylate mixture

Synonym(s):

PTA/TMPTA mixture, Pentaerythritol acrylates mixture

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
6598752
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

300-400 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.483

density

1.18 g/mL at 25 °C

SMILES string

O=C(C=C)OCC(COC(C=C)=O)(COC(C=C)=O)COC(C=C)=O.OCC(COC(C=C)=O)(COC(C=C)=O)COC(C=C)=O.CCC(COC(C=C)=O)(COC(C=C)=O)COC(C=C)=O

InChI

1S/C17H20O8.C15H20O6.C14H18O7/c1-5-13(18)22-9-17(10-23-14(19)6-2,11-24-15(20)7-3)12-25-16(21)8-4;1-5-12(16)19-9-15(8-4,10-20-13(17)6-2)11-21-14(18)7-3;1-4-11(16)19-8-14(7-15,9-20-12(17)5-2)10-21-13(18)6-3/h5-8H,1-4,9-12H2;5-7H,1-3,8-11H2,4H3;4-6,15H,1-3,7

InChI key

JKLIUCYDNJCWHA-UHFFFAOYSA-N

General description

Pentaerythritol tetraacrylate, Pentaerythritoltriacrylate, and Trimethylolpropane triacrylate mixture is a combination of three acrylate monomers widely used for cross-linking. The Pentaerythritol-based acrylate mixtureoffers advantages such as improved mechanical strength, flexibility, chemical resistance, and UV-curing properties. It finds applications in industries such as automotive coatings, electronics, packaging, and 3D bioprinting.

Application

Pentaerythritol tetraacrylate, Pentaerythritol triacrylate, and Trimethylolpropane triacrylate mixture can be used:

  • As a cross-linking agent to synthesize biodegradable poly (1,3-trimethylene carbonate) (PTMC) networks with improved creep resistance and thermal stability. PMTC networks find application in the field of soft tissue engineering.
  • As a monomer precursor to prepare light-curing dental composites via photopolymerization.
  • To fabricate polymer-dispersed liquid crystal(PDLC) films with low driving voltage, moderately high contrast ratio, and fast response time. These PDLC films are utilized in optoelectronic devices such as OLEDs, FET, and solar cells.
  • As a monomer mixture to prepare 3D bioprinting resins viaphotopolymerization.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradable cross-linked poly (1, 3-trimethylene carbonate) networks formed by gamma irradiation under vacuum
Xiliang Liu, et al.
Polymers For Advanced Technologies, 32, 4373-4385 (2021)
Krassimira Yoncheva et al.
International journal of pharmaceutics, 436(1-2), 258-264 (2012-06-23)
Paclitaxel is an antineoplastic drug used against a variety of tumors, but its low aqueous solubility and active removal caused by P-glycoprotein in the intestinal cells hinder its oral administration. In our study, new type of stabilized Pluronic micelles were
Mohammad Mahdi Moein
Talanta, 224, 121794-121794 (2021-01-01)
Since chiral recognition mechanism based on molecularly imprinted polymers immerged, it has assisted countless chemical and electrochemical analytical sample preparation techniques. It has done this by enhancing the enatioseparation abilities of these techniques. The preparation and optimization of chiral molecularly
Hui Huang et al.
Journal of chromatography. A, 1251, 82-90 (2012-07-06)
A mixed-mode monolithic stationary phase was prepared for capillary liquid chromatography (cLC) by in situ copolymerization of 4-vinylphenylboronic acid (VPBA) and pentaerythritol triacrylate (PETA) in a binary porogenic solvent consisting of ethylene glycol/cyclohexanol. The monomer of VPBA functioned as ion-exchange
Recent trends in advanced photoinitiators for vat photopolymerization 3D printing
Yinyin Bao
Macromolecular Rapid Communications, 43, 2200202-2200202 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service