Skip to Content
Merck
All Photos(1)

Key Documents

391913

Sigma-Aldrich

(R)-(+)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid

98%

Synonym(s):

(2R)-3,4-Dihydro-6-hydroxy-2,5,7,8-tetramethyl-2H-1-benzopyran-2-carboxylic acid, (R)-Trolox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]27/D +65°, c = 1.2 in ethanol

mp

162 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

Cc1c(C)c2O[C@](C)(CCc2c(C)c1O)C(O)=O

InChI

1S/C14H18O4/c1-7-8(2)12-10(9(3)11(7)15)5-6-14(4,18-12)13(16)17/h15H,5-6H2,1-4H3,(H,16,17)/t14-/m1/s1

InChI key

GLEVLJDDWXEYCO-CQSZACIVSA-N

General description

(R)-(+)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, a vitamin E analog, is a mushroom tyrosinase inhibitor.

Application

(R)-(+)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid may be used to incorporate antioxidant property while designing anti-stress agents based on γ-aminobutyric acid (GABA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Moreira et al.
Acta poloniae pharmaceutica, 74(2), 688-698 (2018-04-07)
Polyphenol rich extracts obtained from cistus herb (Cistus incanus L.) and pomegranate peel (Punica granatum L.) exhibited significant antioxidant activity in V79 cell culture (Chinese hamster lung fibroblasts) -cistus extract reduced intracellular content of reactive oxygen species (ROS) by 30-40%
Novel compounds designed as antistress agents.
Tsiakitzis, KC, et al.
Journal of Medicinal Chemistry, 52(22), 7315-7318 (2009)
Aleksandra Slezak et al.
Acta poloniae pharmaceutica, 74(2), 670-678 (2018-04-07)
Polyphenol rich extracts obtained from cistus herb (Cistus incanus L.) and pomegranate peels (Punica granatunm L.) added at high concentrations (25-100 pg/mL) to V79 (Chinese hamster lung fibroblast) cell cultures, markedly increased intracellular level of ROS (reactive oxygen species). Also
Shyamal Subramanyam et al.
Methods in enzymology, 600, 201-232 (2018-02-21)
Human RAD51 promotes accurate DNA repair by homologous recombination and is involved in protection and repair of damaged DNA replication forks. The active species of RAD51 and related recombinases in all organisms is a nucleoprotein filament assembled on single-stranded DNA
Inhibitory effects of (S)-and (R)-6-hydroxy-2, 5, 7, 8-tetramethylchroman-2-carboxylic acids on tyrosinase activity.
Yu L.
Journal of Agricultural and Food Chemistry, 51(8), 2344-2347 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service