Skip to Content
Merck
All Photos(2)

Key Documents

381438

Sigma-Aldrich

Benzaldehyde dimethyl acetal

95%

Synonym(s):

α,α-Dimethoxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)2
CAS Number:
Molecular Weight:
152.19
Beilstein:
2044501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

refractive index

n20/D 1.493 (lit.)

bp

87-89 °C/18 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

functional group

acetal
ether
phenyl

SMILES string

COC(OC)c1ccccc1

InChI

1S/C9H12O2/c1-10-9(11-2)8-6-4-3-5-7-8/h3-7,9H,1-2H3

InChI key

HEVMDQBCAHEHDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzaldehyde dimethyl acetal is an organic building block. Pyridinium tosylate-catalyzed acetal exchange reaction between benzaldehyde dimethyl acetal and 6-O-(tert-butyldiphenylsilyl)-1,2-O-isopropylidene-α-D-glucofuranose is reported to afford 3,5-O-benzylidene-1,2-O-isopropylidene-α-D-glucofuranose.[1] The kinetics of the hydrolysis of benzaldehyde dimethyl acetal over amberlite IR-120 has been studied using a circulated batch reactor in dioxane.[2] One-pot tandem conversion of benzaldehydedimethylacetal to trans-1-nitro-2-phenylethylene has been reported.[3]

Application

Benzaldehyde dimethyl acetal is suitable for use in the synthesis of 4,6-dihydroxy sugar, required for the total synthesis of Porphyromonas gingivalis 381 derived lipid A.[4] It may be used in the preparation of 1-O-methyl-2,3-di-O-galloyl-β-D-glucose.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous silica with site-isolated amine and phosphotungstic acid groups: a solid catalyst with tunable antagonistic functions for one-pot tandem reactions.
N Raveendran Shiju et al.
Angewandte Chemie (International ed. in English), 50(41), 9615-9619 (2011-09-16)
K Akerfeldt et al.
Carbohydrate research, 158, 137-145 (1986-12-15)
Pyridinium tosylate-catalyzed acetal exchange between benzaldehyde dimethyl acetal and 6-O-(tert-butyldiphenylsilyl)-1,2-O-isopropylidene-alpha-D-glucofu ranose was investigated as an alternative to the original procedure of Brigl and Grüner (condensation of a D-glucose triol with benzaldehyde under zinc halide catalysis) for synthesis of 3,5-O-benzylidene-1,2-O-isopropylidene-alpha-D-glucofuranose. The
T Ogawa et al.
FEMS immunology and medical microbiology, 28(4), 273-281 (2000-07-13)
A synthetic lipid A of Porphyromonas gingivalis strain 381 (compound PG-381), which is similar to its natural lipid A, demonstrated no or very low endotoxic activities as compared to Escherichia coli-type synthetic lipid A (compound 506). On the other hand
M Toda et al.
Bioscience, biotechnology, and biochemistry, 65(3), 542-547 (2001-05-02)
The clove ellagitannins and their related polygalloyl-glucoses inhibited maltase activity of rat intestinal alpha-glucosidases. The structure-activity relationship study of those galloylglucoses, varying the extent of galloylation on the glucose core, with the ellagitannins, indicated that an increasing number of galloyl
Kinetics of hydrolysis of benzaldehyde dimethyl acetal over Amberlite IR-120.
Altiokka MR and Hosgun HL.
Industrial & Engineering Chemistry Research, 46(4), 1058-1062 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service