Skip to Content
Merck
All Photos(1)

Key Documents

376906

Sigma-Aldrich

D-(+)-3-Phenyllactic acid

98%

Synonym(s):

(R)-2-Hydroxy-3-phenylpropionic acid, (R)-3-Phenyllactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209793
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D +19°, c = 1 in ethanol

mp

122-124 °C (lit.)

functional group

carboxylic acid
hydroxyl
phenyl

SMILES string

O[C@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H10O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)/t8-/m1/s1

InChI key

VOXXWSYKYCBWHO-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block employed in the preparation of statine. Starting material in the preparation of the hypoglycemic agent enlitazone and of 15N-labeled phenylalanine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urban, F.J. et al.
Journal of Heterocyclic Chemistry, 29, 431-431 (1991)
Degerbeck, F. et al.
Journal of the Chemical Society. Perkin Transactions 1, 11-11 (1993)
Journal of Heterocyclic Chemistry, 29, 431-431 (1992)
Wanmeng Mu et al.
Applied microbiology and biotechnology, 95(5), 1155-1163 (2012-07-12)
3-Phenyllactic acid (PLA), which is an organic acid widely existing in honey and lactic acid bacteria fermented food, can be produced by many microorganisms, especially lactic acid bacteria. It was proved as an ideal antimicrobial compound with broad and effective
Demao Li et al.
Se pu = Chinese journal of chromatography, 22(3), 281-283 (2005-02-17)
Various factors affecting the chiral resolution of 3-phenyllactic acid by capillary zone electrophoresis such as the type of cyclodextrin, concentration of the chiral selector, buffer pH, voltage, and temperature were studied and optimized. Using 30 mmol/L hydroxypropyl-beta-cyclodextrin as the chiral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service