Skip to Content
Merck
All Photos(3)

Key Documents

372668

Sigma-Aldrich

Calcium silicate

−200 mesh, 99%

Synonym(s):

Calcium metasilicate, Calcium silicon oxide, Silicon calcium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSiO3
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

reaction suitability

core: calcium

particle size

−200 mesh

SMILES string

[Ca++].[O-][Si]([O-])=O

InChI

1S/Ca.O3Si/c;1-4(2)3/q+2;-2

InChI key

OYACROKNLOSFPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rania Hassan Fahmy
AAPS PharmSciTech, 13(3), 990-1004 (2012-07-19)
Multiparticulate floating drug delivery systems have proven potential as controlled-release gastroretentive drug delivery systems that avoid the "all or none" gastric emptying nature of single-unit floating dosage forms. An objective of the presence investigation was to develop calcium silicate (CaSi)/calcium
S S Salek et al.
Applied microbiology and biotechnology, 97(6), 2743-2752 (2012-11-29)
Silicate minerals can be a source of calcium and alkalinity, enabling CO2 sequestration in the form of carbonates. For this to occur, the mineral needs to be first dissolved in an acidifying process such as the biological process of anaerobic
Bobo Yu et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17465-17476 (2012-11-23)
The aim was to determine the most effective calcium precursor for synthesis of sol-gel hybrids and for improving homogeneity of sol-gel bioactive glasses. Sol-gel derived bioactive calcium silicate glasses are one of the most promising materials for bone regeneration. Inorganic/organic
Emma C Goethals et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 658-666 (2012-10-20)
Polymer nanocapsules have gained an important place as drug delivery vehicles for a myriad of biomedical applications. However, the influence of nanocapsule size, wall thickness, and porosity toward controlling the drug delivery efficiency of nanocapsular systems is not well understood.
Han Guo et al.
International journal of nanomedicine, 7, 3613-3624 (2012-08-01)
The purpose of this study was to synthesize a self-setting bioactive cement by incorporation of wollastonite nanofibers (WNFs) into calcium phosphate cement (CPC). The composition, morphology, setting time, compressive strength, hydrophilicity, and degradation of WNF-doped CPC (wnf-CPC) were investigated. Scanning

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service