Skip to Content
Merck
All Photos(2)

Key Documents

363634

Sigma-Aldrich

Semicarbazide

6 wt. % (on silica gel)

Synonym(s):

Aminourea, Hydrazinecarboxamide, Isosemicarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCONHNH2
CAS Number:
Molecular Weight:
75.07
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

composition

, 6 wt. % (on silica gel)

SMILES string

NNC(N)=O

InChI

1S/CH5N3O/c2-1(5)4-3/h3H2,(H3,2,4,5)

InChI key

DUIOPKIIICUYRZ-UHFFFAOYSA-N

Application

  • Antibacterial activity: Semicarbazide derivatives, specifically thiosemicarbazide, exhibit significant antibacterial properties influenced by substituent position. This underscores its application in pharmaceutical synthesis and microbial resistance studies (Janowska et al., 2024).
  • Solar cell performance: The application of semicarbazide hydrochloride in suppressing deep-level traps significantly improves the efficiency of tin-based perovskite solar cells, highlighting its potential in energy materials research (Jia et al., 2023).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Palcic et al.
Progress in brain research, 106, 41-47 (1995-01-01)
The relationship between the soluble copper topaquinone amine oxidases, the membrane bound semicarbazide-sensitive amine oxidases and lysyl oxidase remains unclear. The stereochemical course of substrate oxidation has been determined for each enzyme type and these studies suggest that SSAO and
Sathya Narayanan Patmanathan et al.
Scientific reports, 6, 25650-25650 (2016-05-11)
Oral squamous cell carcinoma (OSCC) is a lethal disease with a 5-year mortality rate of around 50%. Molecular targeted therapies are not in routine use and novel therapeutic targets are required. Our previous microarray data indicated sphingosine 1-phosphate (S1P) metabolism
Palak J Trivedi et al.
Gut, 67(6), 1135-1145 (2017-04-22)
Primary sclerosing cholangitis (PSC) is the classical hepatobiliary manifestation of IBD. This clinical association is linked pathologically to the recruitment of mucosal T cells to the liver, via vascular adhesion protein (VAP)-1-dependent enzyme activity. Our aim was to examine the
Christian Carpéné et al.
Journal of physiology and biochemistry, 73(3), 475-486 (2016-10-22)
Semicarbazide-sensitive amine oxidase (SSAO), identical to primary amine oxidase or vascular adhesion protein-1, is a membrane enzyme that generates hydrogen peroxide. SSAO is highly expressed at the adipocyte surface, and its plasma levels increase with type 2 diabetes. Since visceral
Jonas M Fuks et al.
PLoS pathogens, 8(12), e1003051-e1003051 (2012-12-14)
During acute infection in human and animal hosts, the obligate intracellular protozoan Toxoplasma gondii infects a variety of cell types, including leukocytes. Poised to respond to invading pathogens, dendritic cells (DC) may also be exploited by T. gondii for spread

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service