Skip to Content
Merck
All Photos(1)

Key Documents

PHR1034

Supelco

Lidocaine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 727
traceable to Ph. Eur. L0595000
traceable to USP 1366002

API family

lidocaine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lidocaine is a drug, which serves as a potential candidate in the treatment of cardiac arrhythmias.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Lidocaine is a local anesthetic drug that can exhibit pronounced antiarrhythmic and anticonvulsant effects. It is known as a central nervous system depressant and shows sedative, analgesic and anticonvulsant properties.[2]

Application

Lidocaine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[3][2][4]
Lidocaine may be used as an internal standard in the determination of perhexiline in human serum samples using gas liquid chromatography (GLC).[5]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Na+ channel blocker; class IB antiarrhythmic.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0142 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

analytical column

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of tolperisone and lidocaine by high performance liquid chromatography
Liawruangrath S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 26(5-6), 865-872 (2001)
A rapid method for the determination of perhexiline in serum using gas-liquid chromatography
CooperHDJ and Turnell CD
Annals of Clinical Biochemistry, 17, 155-158 (1980)
High-performance liquid chromatographic method for the simultaneous determination of lidocaine and its N-dealkylated metabolites in plasma
Nation.LR, et al.
Journal of Chromatography. B, Biomedical Applications, 162, 466-473 (1979)
Stability indicating HPLC method for the estimation of oxycodone and lidocaine in rectal gel
Gebauer MG, et al.
International Journal of Pharmaceutics, 223(1-2), 49-54 (2001)
Development and validation of RP-HPLC method for cetrimonium bromide and lidocaine determination.
Malenovic A, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 60(2), 157-161 (2005)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service