Skip to Content
Merck
All Photos(1)

Documents

359742

Sigma-Aldrich

Trimethylsilyl chlorosulfonate

99%

Synonym(s):

Chlorosulfonic acid trimethylsilyl ester, Trimethylsiloxysulfonyl chloride, Trimethylsilyl chlorosulfate, Trimethylsilyl chlorosulfonate, [Chlorosulfonyloxy(dimethyl)silyl]methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClSO3Si(CH3)3
CAS Number:
Molecular Weight:
188.71
Beilstein:
1763675
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

168-169 °C (lit.)
62-64 °C/10 mmHg (lit.)

mp

−26 °C (lit.)

density

1.225 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OS(Cl)(=O)=O

InChI

1S/C3H9ClO3SSi/c1-9(2,3)7-8(4,5)6/h1-3H3

InChI key

QWQONZVLXJGXHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylsilyl chlorosulfonate is commonly employed as sulfonating agent.

Application

Trimethylsilyl chlorosulfonate may be used:
  • in the sulfonation of small organic molecules
  • in the synthesis of sulfonated polysulfone
  • as sulfonating agent in the preparation of nano-structured organic-inorganic membranes for proton exchange membrane fuel cells (PEMFC) based on methacrylate and epoxy polymeric structures
  • in the preparation of sulfonated polysulfone (sPS)/titanium dioxide (TiO2) composite membranes for use in PEMFCs
  • in the preparation of iodosobenzene sulfate, required for the synthesis of cyclic sulfates

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of sulfonated polysulfone/titanium dioxide composite membranes for proton exchange membrane fuel cells.
Devrim Y, et al.
International Journal of Hydrogen Energy, 34(8), 3467-3475 (2009)
Improved simple synthesis of cyclic sulfates from trimethylsilyl chlorosulfonate.
Bassindale AR, et al.
Canadian Journal of Chemistry, 78(11), 1479-1483 (2000)
Sulfonated polysulfone as promising membranes for polymer electrolyte fuel cells.
Lufrano F, et al.
Journal of Applied Polymer Science, 77(6), 1250-1256 (2000)
Hybrid organic-inorganic nanostructured membranes for high temperature proton exchange membranes fuel cells (PEMFC).
Aparicio M, et al.
Journal of Sol-Gel Science and Technology, 40(2-3), 309-315 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service