P31701
2-Phenylpropionic acid
97%
Synonym(s):
(±)-2-Phenylpropionic acid, (±)-Hydratropic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH(C6H5)CO2H
CAS Number:
Molecular Weight:
150.17
Beilstein:
1863558
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.522 (lit.)
bp
260-262 °C (lit.)
density
1.1 g/mL at 25 °C (lit.)
SMILES string
CC(C(O)=O)c1ccccc1
InChI
1S/C9H10O2/c1-7(9(10)11)8-5-3-2-4-6-8/h2-7H,1H3,(H,10,11)
InChI key
YPGCWEMNNLXISK-UHFFFAOYSA-N
Gene Information
human ... IL8RA(3577)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
298.4 °F
Flash Point(C)
148 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lina Sagert et al.
eLife, 9 (2020-03-14)
Adaptive immunity vitally depends on major histocompatibility complex class I (MHC I) molecules loaded with peptides. Selective loading of peptides onto MHC I, referred to as peptide editing, is catalyzed by tapasin and the tapasin-related TAPBPR. An important catalytic role
Ekaterina Chernevskaya et al.
Critical care (London, England), 24(1), 312-312 (2020-06-10)
High serum levels of certain aromatic microbial metabolites (AMM) are associated with severity and mortality in critically ill patients. Omics-based studies suggest gut dysbiosis and reduced microbiome diversity in critical conditions. However, the landscape of gut microbial metabolites is still
Y Nakamura et al.
Drug metabolism and disposition: the biological fate of chemicals, 15(4), 529-534 (1987-07-01)
Optical isomerization of 2-phenylpropionic acid (hydratropic acid, HTA) was studied in the organs of male rat in vitro. (R)-(-)-HTA was not isomerized by rat liver homogenate even after the addition of CoA, ATP, and Mg2+ to the incubation mixture; however
Chunze Li et al.
The Journal of pharmacology and experimental therapeutics, 305(1), 250-256 (2003-03-22)
Two alternative metabolic pathways, acyl glucuronidation and acyl-CoA formation, are implicated in the generation of reactive acylating metabolites of carboxylic acids. Here, we describe studies that determine the relative importance of these two pathways in the metabolic activation of a
Shengqiang Tong et al.
Journal of chromatography. A, 1281, 79-86 (2013-02-20)
High performance liquid chromatography (HPLC) and high speed counter-current chromatography (HSCCC) were applied and compared in enantioseparation of 2-phenylpropionic acid (2-PPA) when hydroxypropyl-β-cyclodextrin (HP-β-CD) was used as chiral mobile phase additive. For HPLC, the enantioseparation was achieved on ODS C(18)
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service