Skip to Content
Merck
All Photos(1)

Key Documents

358924

Sigma-Aldrich

(+)-O,O′-Diacetyl-L-tartaric anhydride

97%

Synonym(s):

(+)-Diacetyl-L-tartaric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O7
CAS Number:
Molecular Weight:
216.14
Beilstein:
87315
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +59°, c = 6 in acetone

mp

130-135 °C (lit.)

functional group

anhydride
ester

storage temp.

2-8°C

SMILES string

CC(=O)O[C@@H]1[C@@H](OC(C)=O)C(=O)OC1=O

InChI

1S/C8H8O7/c1-3(9)13-5-6(14-4(2)10)8(12)15-7(5)11/h5-6H,1-2H3/t5-,6-/m1/s1

InChI key

XAKITKDHDMPGPW-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-O,O′-Diacetyl-L-tartaric anhydride is an HPLC derivatization reagent for UV/Vis detection. It is mainly employed as a reagent for the chiral derivatization of amino alcohols. It also reacts with alkanoamines in aprotic medium containing trichloroacetic acid and produces tartaric acid monoesters.

Application

(+)-O,O′-Diacetyl-L-tartaric anhydride may be used as a chiral derivatizating agent in the following:
  • determination of enantiomeric vigabatrin in mouse serum samples using ultra-high performance liquid chromatography-quadrupole-time-of-flight mass spectrometry (UHPLC-Q-TOF-M)        
  • determination of trantinterol in rat plasma by ultra performance liquid chromatography–electrospray ionization mass spectrometry (UPLC–MS/MS)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective determination of trantinterol in rat plasma by ultra performance liquid chromatography?electrospray ionization mass spectrometry after derivatization
Yang J, et al.
Talanta, 79(5), 1204-1208 (2009)
Determination of enantiomeric vigabatrin by derivatization with diacetyl-l-tartaric anhydride followed by ultra-high performance liquid chromatography-quadrupole-time-of-flight mass spectrometry
Zhao J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1040, 199-207 (2017)
D R Brocks et al.
Journal of chromatography, 581(1), 83-92 (1992-10-02)
(+/-)-Hydroxychloroquine (HCQ) is an antimalarial and anti-arthritic drug which is administered as the racemate. An accurate, precise and sensitive high-performance liquid chromatographic assay was developed for the determination of HCQ enantiomers in samples from human plasma, serum, whole blood, and
William M Oldham et al.
Bio-protocol, 6(16) (2017-06-03)
Two enantiomers of 2-hydroxyglutarate (2HG), L (L2HG) and D (D2HG), are metabolites of unknown function in mammalian cells that were initially associated with separate and rare inborn errors of metabolism resulting in increased urinary excretion of 2HG linked to neurological
D R Brocks et al.
Journal of pharmaceutical and biomedical analysis, 13(7), 911-918 (1995-06-01)
A stereospecific liquid chromatographic (LC) assay was developed for the quantification of the antimalarial drug, halofantrine, in human plasma. Following protein precipitation with acetonitrile, the enantiomers of halofantrine were extracted from human plasma using ammonium hydroxide and tert-butyl methyl ether-hexane.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service