Skip to Content
Merck
All Photos(1)

Key Documents

348848

Sigma-Aldrich

Titanium

foil, thickness 0.025 mm, 99.98% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
EC Number:
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.98% trace metals basis

form

foil

autoignition temp.

860 °F

resistivity

42.0 μΩ-cm, 20°C

thickness

0.025 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

Ti has low thermal and electrical conductivity. It is highly corrosion-resistant and has a high strength to weight ratio. A few angstroms thick layer of titania on the surface of Ti products makes its corrosion resistant.5 Ti foils could be employed as a substrate to grow arrays of hematite nanorods by hydrothermal method.[1] A study reports pressure less sintering of SiC pieces and single crystals to Ti foils at 1500oC.[2] Ti foil may be applied as an interlayer, diffusion bonded during the self joining of Si3N4. Si3N4/Ti-foil/Si3N4.[3]

Application

  • Medical Devices: Due to its biocompatibility, it is widely used in medical applications, such as in joint replacement implants, dental implants, and surgical instruments (Britannica).
  • Corrosion Resistance: Its resistance to corrosion by both water and chemical media leads to its use in chemical processing industries for equipment like heat exchangers and reactors (Royal Society of Chemistry).

Quantity

280 mg = 50 × 50 mm; 1.1 g = 100 × 100 mm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Joining of silicon nitride with a titanium foil interlayer
Lemus J and Drew RAL
Materials Science & Engineering. A, Structural Materials : Properties, Microstructure and Processing, 352, 169-178 (2003)
Bonding mechanism between silicon carbide and thin foils of reactive metals
Morozumi S, et al.
J. Mater. Sci., 20(11), 3976-3982 (1985)
Large-Scale Porous Hematite Nanorod Arrays: Direct Growth on Titanium Foil and Reversible Lithium Storage.
Song Y et al.
The Journal of Physical Chemistry C, 114(49), 21158-21164 (2010)
Jinho Shin et al.
Journal of nanoscience and nanotechnology, 13(8), 5807-5810 (2013-07-26)
In this study, hydroxyapatite (HA) was coated on anodized titanium (Ti) surfaces through radio frequency magnetron sputtering in order to improve biological response of the titanium surface. All the samples were blasted with resorbable blasting media (RBM). RBM-blasted Ti surface
Study of a TiO2 photocatalytic coating for use in plasma catalysis.
K Van Wesenbeeck et al.
Communications in agricultural and applied biological sciences, 78(1), 227-233 (2013-07-24)

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service