Skip to Content
Merck
All Photos(1)

Key Documents

345458

Sigma-Aldrich

Boron trichloride solution

1.0 M in p-xylene

Synonym(s):

Boron chloride, Trichloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.03 (vs air)

vapor pressure

1 mmHg ( 12.7 °C)

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

1.0 M in p-xylene

density

0.901 g/mL at 25 °C

SMILES string

ClB(Cl)Cl

InChI

1S/BCl3/c2-1(3)4

InChI key

FAQYAMRNWDIXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Desvergnes et al.
Organic letters, 10(14), 2967-2970 (2008-06-24)
A general method to prepare a new class of carbohydrate-derived, enantiomerically pure polyhydroxypyrroline N-oxides from their alkoxy (protected) derivatives is presented. Boron trichloride is shown to cleave efficiently benzyl ethers and ketals without affecting the imine N-oxide functionality of nitrones.
Lei Zhang et al.
Journal of combinatorial chemistry, 8(3), 361-367 (2006-05-09)
A novel and efficient microwave-assisted, BCl(3) mediated coupling reaction to synthesize o-(hydroxyaryl)(aryl)methanone structures from phenols and acyl chlorides is described. This reaction was further incorporated into a two-step synthesis of biologically interesting xanthones.
Stephen Sproules et al.
Inorganic chemistry, 50(10), 4503-4514 (2011-04-19)
Sky-blue Tp*WOCl(2) has been synthesized from the high-yielding reaction of Tp*WO(2)Cl with boron trichloride in refluxing toluene. Dark-red Tp*WOI(2) was prepared via thermal decarbonylation followed by aerial oxidation of Tp*WI(CO)(3) in acetonitrile. From these precursors, an extensive series of mononuclear
Warwick J Belcher et al.
Dalton transactions (Cambridge, England : 2003), (12)(12), 1602-1614 (2008-03-13)
The reactions of boron halides with free base porphyrins under conditions where partial hydrolysis of the boron halides can occur give diboron porphyrin complexes containing BOB moieties in which each boron is bonded to two porphyrin nitrogen atoms. BF(3).OEt(2) with
Robert Winker et al.
International archives of occupational and environmental health, 81(4), 423-428 (2007-07-27)
To examine whether semiconductor workers exposed to complex mixtures of chemical waste show an increase in genotoxic effects, and, if so, whether occupational safety measures protect these workers. To assess chemical exposure in the workplace, air monitoring of boron trifluoride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service