Skip to Content
Merck
All Photos(2)

Key Documents

178934

Sigma-Aldrich

Boron trichloride solution

1.0 M in methylene chloride

Synonym(s):

Boron chloride, Trichloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

29.72 psi ( 55 °C)
9.05 psi ( 20 °C)

Quality Level

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

1.0 M in methylene chloride

density

1.326 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClB(Cl)Cl

InChI

1S/BCl3/c2-1(3)4

InChI key

FAQYAMRNWDIXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Boron trichloride is a boron halide. It participates in the hydroboration of alkenes. It forms strongly polarized donor-acceptor complexes with trimethyl- and triethylsilyl triflate. Boron trichloride assisted reaction between acid chlorides and alkynyltrifluoroborate salts have been reported to form ynones.

Boron trichloride solution is used as a Lewis acid catalyst in the synthesis of BN-phenanthrenes.

Application

Boron trichloride may be used in the synthesis of Cl-BODIPYs (BODIPY - Boron-dipyrromethene) and tris(dimethylamino)borane. It may be used in the synthesis of sceptrins and nakamuric acid.
For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of highly electrically conductive and electrochemically stable porous boron-doped carbon microspheres
Bose, Anima B and Yang, et al.
SN Applied Sciences, 1, 1-8 (2019)
A New Member of the BN-Phenanthrene Family: Understanding the Role of the B?N Bond Position
Abengo?zar, Alberto and Sucunza
The Journal of Organic Chemistry, 84, 7113-7122 (2019)
Organometallic chemistry. 21. Silyl trifluoromethanesulfonate (triflate)-boron trichloride (tribromide) complexes.
Olah GA, et al.
Organometallics, 3(9), 1337-1340 (1984)
Hydroboration with boron halides and trialkylsilanes.
Soundararajan R and Matteson DS.
The Journal of Organic Chemistry, 55(8), 2274-2275 (1990)
Bis (pinacolato) diboron.
Ishiyama T, et al.
Organometallic Syntheses, 77, 176-185 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service