Skip to Content
Merck
All Photos(1)

Key Documents

345083

Sigma-Aldrich

Copper bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

99%

Synonym(s):

2,2,6,6-Tetramethyl-3,5-heptanedione copper(II) derivative, Cu(TMHD)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(OCC(CH3)3CHCOC(CH3)3)2
CAS Number:
Molecular Weight:
430.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99%

form

solid

reaction suitability

core: copper

mp

198 °C (dec.) (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(/O[Cu]O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/2C11H20O2.Cu/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;/h2*7,12H,1-6H3;/q;;+2/p-2/b2*8-7-;

InChI key

VCIKJQZFNVXWPF-ATMONBRVSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 29 Copper

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service