All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H10N6
CAS Number:
Molecular Weight:
238.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
182-184 °C (lit.)
SMILES string
Nn1c(nnc1-c2ccccn2)-c3ccccn3
InChI
1S/C12H10N6/c13-18-11(9-5-1-3-7-14-9)16-17-12(18)10-6-2-4-8-15-10/h1-8H,13H2
InChI key
VHRKXLULSZZZQT-UHFFFAOYSA-N
General description
4-Amino-3,5-di-2-pyridyl-4H-1,2,4-triazole reacts with iron(II) sulfate and pentacyanidonitrosylferrate(II) or hexacyanidoplatinate(IV) to yield one-dimensional iron(II) spin-crossover compounds. 4-Amino-3,5-di-2-pyridyl-4H-1,2,4-triazole (abpt) reacts with CoCl2·6H2O and KSCN to yield mononuclear complex named [CoSCN(abpt)] and 2D inorganic coordination framework K2[Co3(OH)2(SO4)3(H2O)2].
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Radovan Herchel et al.
Inorganic chemistry, 50(24), 12390-12392 (2011-11-22)
The reactions of iron(II) sulfate, 4-amino-3,5-di-2-pyridyl-4H-1,2,4-triazole (abpt), and pentacyanidonitrosylferrate(II) or hexacyanidoplatinate(IV) resulted in the formation of one-dimensional iron(II) spin-crossover compounds [Fe(abpt)(2)(μ-Fe(CN)(5)(NO))](n) (1) and [Fe(abpt)(2)(μ-Pt(CN)(6))](n) (2) with the spin-transition critical temperature near or above room temperature accompanied by thermochromism. Furthermore, it
Hydrothermal Synthesis and Crystal Structure of 2D K2[Co3(OH)2(SO4)3 (H2O)2] Coordination Polymer with the In-Situ Formation of SO4 (2-).
Chen Z-Y and Peng M-X.
Chinese Journal of Inorganic Chemistry / Wu Ji Hua Xue Xue Bao, 23(12)2007, 2091-2096 null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service