Skip to Content
Merck
All Photos(1)

Key Documents

338737

Sigma-Aldrich

4-Bromo-3-nitroanisole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(NO2)OCH3
CAS Number:
Molecular Weight:
232.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

153-154 °C/13 mmHg (lit.)

mp

32-34 °C (lit.)

SMILES string

COc1ccc(Br)c(c1)[N+]([O-])=O

InChI

1S/C7H6BrNO3/c1-12-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3

InChI key

KCOBIBRGPCFIGF-UHFFFAOYSA-N

General description

FT-IR and FT-Raman spectra of 4-bromo-3-nitroanisole have been studied.

Application

4-Bromo-3-nitroanisole was used in the synthesis of:
  • 4-bromo-5-methoxyaniline
  • 2-(4-methoxy-2-nitrophenylsulfanyl)benzoic acid
  • 4-(1-diethylamino-4-pentylamino)-3-nitrochlorobenzene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of soluble polyarylenes containing alternating 4, 4'-(1, 1'-binaphthyl) and 4, 4'-(3, 3'-diphenyl) biphenyl structural units.
Percec V, et al.
Polymer Bull., 29(3-4), 271-276 (1992)
V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 284-298 (2013-02-19)
The FT-IR and FT-Raman spectra of 4-bromo-3-nitroanisole (BNA) molecule have been recorded in the region 4000-400 cm(-1) and 3500-100 cm(-1), respectively. Optimized geometrical structure, harmonic vibrational frequencies, intensities, reduced mass, force constants and depolarization ratio have been computed by ab
Some derivatives of 3-amino-4-(1-diethylamino-4-pentylamino)-chlorobenzene.
R L McKEE et al.
Journal of the American Chemical Society, 68(10), 1904-1904 (1946-10-01)
A A Wilson et al.
Journal of medicinal chemistry, 43(16), 3103-3110 (2000-08-24)
A series of four 2-(phenylthio)araalkylamines have been radiolabeled with (11)C and evaluated as potential radiotracers for imaging the serotonin transporter (SERT) by positron emission tomography (PET). All four candidates display high affinity for SERT and low affinity for the dopamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service