Skip to Content
Merck
All Photos(3)

Key Documents

333379

Sigma-Aldrich

Potassium ethoxide

95%

Synonym(s):

Potassium ethanolate, Potassium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOC2H5
CAS Number:
Molecular Weight:
84.16
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

SMILES string

[K+].CC[O-]

InChI

1S/C2H5O.K/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

RPDAUEIUDPHABB-UHFFFAOYSA-N

Application

Potassium ethoxide can be used:
  • As a catalyst for transesterification reactions for the synthesis of biodiesel.
  • To synthesize nitrogen doped carbon nano-sheets to be used as an anode in sodium ion batteries.
  • To promote the arylation reaction of unactivated arenes to synthesize biaryl compounds.
  • As a precursor for the synthesis of thin films of (Na0.5K0.5)NbO3 (NKN) and K3Li2Nb5O15 (KLN).
  • For ethoxylation of p-chloronitrobenzene in the presence of phase transfer catalysts.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanzhan Zeng et al.
ACS nano, 14(1), 611-619 (2020-01-01)
The rapid development of modern electrical equipment toward miniaturization and high power puts forward stringent requirements to the mechanical reliability, dielectric property, and heat resistance of electrical insulating materials. Simultaneous integration of all these properties for mica-based materials remains unresolved.
Continuous production of ethanol-based biodiesel under subcritical conditions employing trace amount of homogeneous catalysts
Liu J, et al.
Fuel: The Science and Technology of Fuel and Energy, 193, 187-196 (2017)
A novel aqueous solution method for K3Li2Nb5O15 film and powder
Cheng Z, et al.
Journal of Crystal Growth, 307(2), 353-357 (2007)
Fabrication of highly oriented lead-free (Na, K) NbO3 thin films at low temperature by sol-gel process
Tanaka K, et al.
Journal of Crystal Growth, 294(2), 209-213 (2006)
Nitrogen doped holey carbon nano-sheets as anodes in sodium ion battery
Vadahanambi S, et al.
Royal Society of Chemistry Advances, 6(44), 38112-38116 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service