Skip to Content
Merck
All Photos(4)

Key Documents

332593

Sigma-Aldrich

(2-Hydroxypropyl)-β-cyclodextrin

average Mw ~1,380

Synonym(s):

(2-Hydroxypropyl)-beta-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

powder

optical activity

[α]26/D +139°, c = 1 in H2O

mol wt

average Mw ~1,380

extent of labeling

0.6 molar substitution

mp

278 °C (dec.)

solubility

water: 180 g/L

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(2-Hydroxypropyl)-β-cyclodextrin (HP-β-CD) is a beta-cyclodextrin and hydroxyalkyl derivative and is widely used as a chiral solvent, phase transfer catalyst, and host molecule in organic synthesis. HP-β-CD can form inclusion complexes with guest molecules, which can help enhance the solubility, stability, and bioavailability of the guest molecule. It is an alternative to α-, β- and γ-cyclodextrin and exhibits a higher water solubility. HP-β-CD is commonly used as a drug solubilizer.

Application

HP-β-CD can be used:
  • As an additive in the separation of amino acids and lactic acids based on the coordination interactions between Cu2+ ions and analytes.
  • As a chiral selector in capillary electrophoresis, enabling the robust separation of diastereomeric derivatives of aspartic acid for analysis.
  • As a reagent to improve water solubility and anti-proliferative activity of pyrazolo[3,4-d] pyrimidines.
  • As formulation vehicle. It increases water solubility of the drug for successful delivery of medical agents to a biological system.
  • To increase the solubility of ropivacaine (RVC) upon complexation, which in turn enhances the pharmacological activity of RVC.
  • To increase the aqueous solubility of naproxen (NAP) in presence of polyvinylpyrrolidone (PVP).
  • As a template in the synthesis of hollow spherical copper sulfide nanoparticle assemblies.
The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

>752.0 °F

Flash Point(C)

> 400 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of polyvinylpyrrolidone on naproxen complexation with hydroxypropyl- β -cyclodextrin
Mura P, et al.
European Journal of Pharmaceutical Sciences, 13(2), 187-194 (2001)
The fabrication of hollow spherical copper sulfide nanoparticle assemblies with 2-hydroxypropyl- β -cyclodextrin as a template under sonication
Xu J-Z, et al.
Ultrasonics Sonochemistry, 13(5), 451-454 (2006)
Evaluation of enantiomeric purity of magnesium-L-aspartate dihydrate
Oliver W,et al.
Journal of Pharmaceutical and Biomedical Analysis, 102, 100-109 (2015)
Magdalena Koziczak-Holbro et al.
The Journal of pharmacology and experimental therapeutics, 369(2), 188-199 (2019-03-02)
The anabolic effects of β2-adrenoceptor (β2-AR) agonists on skeletal muscle have been demonstrated in various species. However, the clinical use of β2-AR agonists for skeletal muscle wasting conditions has been limited by their undesired cardiovascular effects. Here, we describe the
2-Hydroxypropyl-beta-cyclodextrin strongly improves water solubility and anti-proliferative activity of pyrazolo [3, 4-d] pyrimidines Src-Abl dual inhibitors
Elena D,et al.
European Journal of Medicinal Chemistry, 45, 5958-5964 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service