Skip to Content
Merck
All Photos(1)

Documents

W402826

Sigma-Aldrich

β-Cyclodextrin

produced by Wacker Chemie AG, Burghausen, Germany

Synonym(s):

Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
FEMA Number:
4028
Beilstein:
78623
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

grade

produced by Wacker Chemie AG, Burghausen, Germany

description

derived from GMO corn

form

powder

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

color

white

mp

290-300 °C (dec.) (lit.)

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Cyclodextrin is a cyclic oligosaccharide with a cage-like supramolecular structure. The internal cavity is apolar and can form inclusion complexes with many compounds such as fats, flavors, and colors. This molecular complexation ability of β-CD is used in the food industry for flavor protection or flavor delivery and in the cosmetic industry for the controlled release of fragrances.

Application


  • Modulating the ion-transfer electrochemistry of perfluorooctanoate with serum albumin and beta-cyclodextrin.: This article discusses the modification of the electrochemical properties of perfluorooctanoate through interactions with beta-cyclodextrin and serum albumin, illustrating the utility of beta-cyclodextrin in environmental cleanup technologies and biochemical sensing (Lamichhane & Arrigan, 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclodextrins and their uses: a review.
Del Valle EM.
Biotechnology Advances, 39(9), 1033-1046 (2004)
Industrial applications of cyclodextrins.
Hedges AR.
Process Biochemistry (Oxford, United Kingdom), 98(5), 2035-2044 (1998)
Biotechnological applications of cyclodextrins.
Singh M, et al.
BioTechnology: An Indian Journal, 20(5), 341-359 (2002)
A review on the use of cyclodextrins in foods.
Astray G, et al.
Food Hydrocolloids, 23(7), 1631-1640 (2009)
Characterization of beta-cyclodextrin inclusion complexes containing essential oils (trans-cinnamaldehyde, eugenol, cinnamon bark, and clove bud extracts) for antimicrobial delivery applications.
Hill LE, et al.
LWT--Food Science and Technology, 51(1), 86-93 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service