Skip to Content
Merck
All Photos(1)

Key Documents

324787

Sigma-Aldrich

2,2,2-Trifluoroethanesulfonyl chloride

99%

Synonym(s):

Tresyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2SO2Cl
CAS Number:
Molecular Weight:
182.55
Beilstein:
1860983
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.388 (lit.)

bp

140-141 °C (lit.)
64-67 °C/45 mmHg (lit.)

density

1.651 g/mL at 25 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)CS(Cl)(=O)=O

InChI

1S/C2H2ClF3O2S/c3-9(7,8)1-2(4,5)6/h1H2

InChI key

CXCHEKCRJQRVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tohru Hayakawa et al.
Journal of biomedical materials research. Part A, 67(2), 684-688 (2003-10-21)
The aim of this study was to bind fibronectin directly to a titanium surface treated with tresyl chloride (2,2,2-trifluoroethanesulfonyl chloride) for the development of a strong connection of a dental implant to subepithelial connective tissues and/or peri-implant epithelia. Basic terminal
Kristina Jonsson-Schmunk et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(5), 758-770 (2016-02-13)
Landmark studies describing the effect of microbial infection on the expression and activity of hepatic CYP3A used bacterial lipopolysaccharide as a model antigen. Our efforts to determine whether these findings were translatable to viral infections led us to observations suggesting
Tresyl chloride-activated supports for enzyme immobilization.
K Nilsson et al.
Methods in enzymology, 135, 65-78 (1987-01-01)
Q Yang et al.
Analytical biochemistry, 268(2), 354-362 (1999-03-17)
For immobilized (proteo)liposome chromatography, unilamellar liposomes were covalently bound within gel beads that had been activated by CNBr, N-hydroxysuccinimide, tresyl, or chloroformate. Liposomes composed of phosphatidylcholine (PC) and 2 mol% of amino-containing lipid (phosphatidylethanolamine-caproylamine) were immobilized in the activated gels
D A Puleo
Biomaterials, 17(2), 217-222 (1996-01-01)
Because of the limited mechanical properties of tissue substitutes formed by culturing cells on polymeric scaffolds, other approaches to tissue engineering must be explored for applications that require complete and immediate ability to bear weight, e.g. total joint replacements. Biochemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service