Skip to Content
Merck
All Photos(4)

Documents

323446

Sigma-Aldrich

Ammonium tetrathiomolybdate

99.97% trace metals basis

Synonym(s):

Ammonium thiomolybdate, Diammonium tetrathiomolybdate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2MoS4
CAS Number:
Molecular Weight:
260.28
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.97% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤350.0 ppm Trace Metal Analysis

mp

>300 °C (lit.)

SMILES string

[NH4+].[NH4+].[S-][Mo]([S-])(=S)=S

InChI

1S/Mo.2H3N.4S/h;2*1H3;;;;/q;;;;;2*-1/p+2

InChI key

PQNOIAHNKHBLRN-UHFFFAOYSA-P

Looking for similar products? Visit Product Comparison Guide

Application

Ring opening of aziridines leading to sulfides and disulfides.
This classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.

Biochem/physiol Actions

Acts as a chelator in vivo to reduce copper levels. Tetrathiomolybdate interferes with angiogenesis and reduces tumor growth. Its mechanism may involve inhibition of NF-κB and downstream cytokines.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Cao et al.
Scientific reports, 7(1), 16048-16048 (2017-11-24)
Amorphous molybdenum sulfide/carbon black (MoS
Yu Cheng et al.
Molecular pharmaceutics, 16(8), 3489-3501 (2019-06-28)
Recently, multifunctional clearable inorganic theranostic nanoparticles have been attracting more and more attention. Protein-based nanoparticles can be cleared by the hepatobiliary system efficiently. In this work, ultrasmall gadolinium oxide (Gd2O3) nanoparticles, which possess the advantage of high longitudinal relaxation rate
Devarajulu Sureshkumar et al.
The Journal of organic chemistry, 72(6), 2106-2117 (2007-02-24)
A comprehensive study of a general and effective one-step procedure for the synthesis of beta-sulfonamidodisulfides directly from N-tosyl aziridines in a regio- and stereospecific manner under neutral conditions without the use of any Lewis acid or base has been reported.
George J Brewer
Current cancer drug targets, 5(3), 195-202 (2005-05-17)
Tetrathiomolybdate (TM) is a novel anticopper agent under development for use in Wilson's disease. It acts by forming a stable tripartite complex with serum albumin and copper, rendering the complexed copper unavailable for cellular uptake. TM is a very potent
Yu Lei et al.
Science advances, 6(32), eabc4250-eabc4250 (2020-08-22)
Two-dimensional transition metal dichalcogenides (TMDs) emerged as a promising platform to construct sensitive biosensors. We report an ultrasensitive electrochemical dopamine sensor based on manganese-doped MoS2 synthesized via a scalable two-step approach (with Mn ~2.15 atomic %). Selective dopamine detection is

Articles

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service