290912
1,4-Bis(trifluoromethyl)benzene
98%
Synonym(s):
α,α,α,α′,α′,α′-Hexafluoro-p-xylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H4(CF3)2
CAS Number:
Molecular Weight:
214.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.379 (lit.)
bp
116 °C (lit.)
density
1.381 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
FC(F)(F)c1ccc(cc1)C(F)(F)F
InChI
1S/C8H4F6/c9-7(10,11)5-1-2-6(4-3-5)8(12,13)14/h1-4H
InChI key
PDCBZHHORLHNCZ-UHFFFAOYSA-N
Related Categories
General description
The infrared absorption of liquid 1,4-bis(trifluoromethyl) benzene has been studied using LiF, NaCl and KBr prisms.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
71.6 °F - closed cup
Flash Point(C)
22 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Vibrational Spectra of Fluorinated Aromatics. VIII. 1, 4-Bis (trifluoromethyl) benzene.
Ferguson EE, et al.
J. Chem. Phys. , 21(10), 1731-1735 (1953)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service