Skip to Content
Merck
All Photos(1)

Key Documents

290890

Sigma-Aldrich

Dibutyltin diacetate

technical grade

Synonym(s):

Diacetoxydibutyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2Sn(OCOCH3)2
CAS Number:
Molecular Weight:
351.03
Beilstein:
4136035
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

12 (vs air)

vapor pressure

1.3 mmHg ( 25 °C)

refractive index

n20/D 1.471 (lit.)

bp

142-145 °C/10 mmHg (lit.)

mp

7-10 °C (lit.)

density

1.32 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(OC(C)=O)OC(C)=O

InChI

1S/2C4H9.2C2H4O2.Sn/c2*1-3-4-2;2*1-2(3)4;/h2*1,3-4H2,2H3;2*1H3,(H,3,4);/q;;;;+2/p-2

InChI key

JJLKTTCRRLHVGL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyltin diacetate is an organotin compound that is commonly used as a catalyst in organic synthesis.

Application

Dibutyltin diacetate can be used as a catalyst to synthesize:
  • Wittenberger tetrazole derivatives by reacting with various nitriles in the presence of trimethylsilyl azide.
  • Carbamate derivatives from various isocyanates and alcohols in the presence of dichloromethane as solvent.
  • Urea derivatives from aryl isocyanates and various amines in the presence of 4-methylmorpholine.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 1 Oral

Target Organs

thymus gland, thymus gland,Immune system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.9 °F - Pensky-Martens closed cup

Flash Point(C)

150.5 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Blaser et al.
Journal of medicinal chemistry, 55(1), 312-326 (2011-12-14)
Analogues of clinical tuberculosis drug (6S)-2-nitro-6-{[4-(trifluoromethoxy)benzyl]oxy}-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine (PA-824), in which the OCH(2) linkage was replaced with amide, carbamate, and urea functionality, were investigated as an alternative approach to address oxidative metabolism, reduce lipophilicity, and improve aqueous solubility. Several soluble monoaryl examples
TMSN3-Bu2Sn (OAc) 2: A modified and mild reagent system for Wittenberger tetrazole-synthesis
Yoneyama H, et al.
Tetrahedron Letters, 61, 151517-151517 (2020)
Yun-Chae Jung et al.
Scientific reports, 7(1), 17482-17482 (2017-12-14)
Amorphous poly(ethylene ether carbonate) (PEEC), which is a copolymer of ethylene oxide and ethylene carbonate, was synthesized by ring-opening polymerization of ethylene carbonate. This route overcame the common issue of low conductivity of poly(ethylene oxide)(PEO)-based solid polymer electrolytes at low
T Noda et al.
Archives of environmental contamination and toxicology, 23(2), 216-222 (1992-08-01)
Teratological tests were conducted on di-n-butyltin diacetate (DBTA), and n-butyltin trichloride (MBTC). Pregnant Wistar rats were treated orally with DBTA at doses of 0, 1.7, 5.0, 10.0, and 15.0 mg/kg/day or with MBTC at doses of 0, 50, 100, 200
B B Harsanyi et al.
Journal of dental research, 70(6), 991-996 (1991-06-01)
The hamster cheek pouch provides a suitable model system for the mucous-membrane irritation testing of dental materials. Poor retention of materials or difficulties in histopathological interpretation caused by surgical artifacts have been reported in published techniques. We describe a new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service