Skip to Content
Merck
All Photos(1)

Key Documents

290831

Sigma-Aldrich

Bis(2-ethylhexyl) sebacate

technical grade, 90%

Synonym(s):

Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
426.67
Beilstein:
1806504
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

90%

refractive index

n20/D 1.450 (lit.)

bp

212 °C/1 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) sebacate, or dioctyl sebacate (DOS), is a non-polar plasticizer that has two ester groups that bond with cationic compounds. It is mainly used in the formation of thin plasticized polyvinyl chloride (PVC) based membranes.

Application

DOS can be used as an aerosol material that can be used to investigate the filtration efficiency of polyvinylidene/silicon oxide (PVDF/SiO2) based fibrous membranes. It may be blended with PVC to form membranes that can be deposited on silver (Ag) based composites which can be used in the fabrication of ion-selective electrodes for chemical analysis. DOS may also be used in combination with PVC, chloride ionophore III, and tridodecylmethylammonium chloride for the development of chloride sensing films.

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jain, A.K. et al.
Electrochimica Acta, 51(12), 2006-2006 (2006)
Calibration free solid contact electrodes with two PVC based membranes.
Bartoszewicz B, et al.
Sensors and Actuators B, Chemical, 274(2), 268-273 (2018)
Developing ultra-high gas permeance PVDF hollow fibers for air filtration applications.
Wang L, et al.
Separation and Purification Technology, 205(2), 184-195 (2018)
Investigation of Sensitivities and Drift Effects of the Arrayed Flexible Chloride Sensor Based on RuO2/GO at Different Temperatures.
Tseng S, et al.
Sensors, 18(2), 632-632 (2018)
Zielinska, R.,
Analytical Chemistry, 451, 243-243 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service