Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bis(2-ethylhexyl) sebacate
Synonym(s):
Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate, Bis(2-ethylhexyl) sebacate
Linear Formula:
[-(CH
2
)
4
CO
2
CH
2
CH(C
2
H
5
)(CH
2
)
3
CH
3
]
2
CAS Number:
122-62-3
Molecular Weight:
426.67
EC Number:
204-558-8
Products
Product Category
building blocks (1)
esters (1)
Brand
Sigma-Aldrich (3)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Purum (1)
Technical (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
84822
Bis(2-ethylhexyl) sebacate,
≥97.0% (GC)
Expand
View Pricing
290831
Bis(2-ethylhexyl) sebacate,
technical grade, 90%
Expand
View Pricing
8.14576
Bis(2-ethylhexyl) sebacate,
for synthesis
Expand
View Pricing
69027
Bis(2-ethylhexyl) sebacate,
analytical standard
Expand
View Pricing
84818
Bis(2-ethylhexyl) sebacate,
Selectophore
™
, ≥97.0%
Expand
View Pricing