Skip to Content
Merck
All Photos(3)

Key Documents

281972

Sigma-Aldrich

3-Chloro-2-methyl-1-propene

contains ~250 ppm Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) as stabilizer, 98%

Synonym(s):

β-Methallyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)CH2Cl
CAS Number:
Molecular Weight:
90.55
Beilstein:
878160
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050134
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.12 (vs air)

Quality Level

vapor pressure

102 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

899 °F

contains

~250 ppm Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) as stabilizer

expl. lim.

9.3 %

refractive index

n20/D 1.427 (lit.)

bp

71-72 °C (lit.)

mp

−80 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CC(=C)CCl

InChI

1S/C4H7Cl/c1-4(2)3-5/h1,3H2,2H3

InChI key

OHXAOPZTJOUYKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Chloro-2-methyl-1-propene is also known as 2-methallyl chloride, methyl allyl chloride and isobutenyl chloride.

Application

3-Chloro-2-methyl-1-propene is a widely used insecticide and a chemical intermediate. It has been used as reactant in the synthesis of cyclobutanone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

National Toxicology Program technical report series, 300, 1-196 (1986-06-01)
Toxicology and carcinogenesis studies of technical-grade 3-chloro-2-methylpropene (containing 5% dimethylvinyl chloride), a widely used insecticide and a chemical intermediate, were performed on F344/N rats and B6C3F1 mice. In the 13-week studies, 50%-100% mortality occurred in groups of male and female
Tze Loon Neoh et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(10), 1690-1702 (2017-05-18)
The methodology to electrospin polystyrene (PS) fibres functionalised with the inclusion complex between 1-methylcyclopropene (1-MCP) and α-cyclodextrin (α-CD) has been developed successfully. Due to limited availability, α-CD crystals instead of 1-MCP/α-CD complex were suspended in the electrospinning (ES) solutions to
3-Chloro-2-methylpropene.
Report on carcinogens : carcinogen profiles, 11, III57-III58 (2004-01-01)
3-Chloro-2-methylpropene.
Report on carcinogens : carcinogen profiles, 10, 57-58 (2004-08-24)
3-Chloro-2-methylpropene.
IARC monographs on the evaluation of carcinogenic risks to humans, 63, 325-333 (1995-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service