279730
Tris(dimethylamino)borane
97%
Synonym(s):
Hexamethylboranetriamine, NSC 379532
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
B(N(CH3)2)3
CAS Number:
Molecular Weight:
143.04
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
reaction suitability
reagent type: reductant
refractive index
n20/D 1.446 (lit.)
bp
147-148 °C (lit.)
mp
−16 °C (lit.)
density
0.817 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)B(N(C)C)N(C)C
InChI
1S/C6H18BN3/c1-8(2)7(9(3)4)10(5)6/h1-6H3
InChI key
SOLWORTYZPSMAK-UHFFFAOYSA-N
General description
Tris(dimethylamino)borane is an organoborane generally used as a macrolactamization reagent. It can also be used to prepare tris(alkylamino)borane analogs by transamination of primary and secondary amines.
Application
Reactant involved in:
- B-C-N hybrid films deposited on Ni
- Reduction reactions
- Syntehsis of lithium borom imidazolate framework polymeric complexes
- One-pot oxidative amination
- Synthesis of boron salts and tripodal borate ligands
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-5.8 °F - closed cup
Flash Point(C)
-21 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tris(dimethylamino)borane
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2008)
Journal of Organometallic Chemistry, 303, 324-324 (1986)
Tetrahedron Letters, 32, 639-639 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service