Skip to Content
Merck
Tributyl(1-ethoxyvinyl)tin 97%

Tributyl(1-ethoxyvinyl)tin

Tributyl(1-ethoxyvinyl)tin 97%
Linear Formula:
[CH3(CH2)3]3SnC(OC2H5)=CH2
CAS Number:
Molecular Weight:
361.15

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing