Skip to Content
Merck
All Photos(3)

Documents

269093

Sigma-Aldrich

1,3-Benzenedithiol

99%

Synonym(s):

Benzene-1,3-dithiol, 1,3-Dimercaptobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(SH)2
CAS Number:
Molecular Weight:
142.24
Beilstein:
1634412
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

bp

128-129 °C/16 mmHg (lit.)

mp

24-25 °C (lit.)

SMILES string

SC1=CC(S)=CC=C1

InChI

1S/C6H6S2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

ZWOASCVFHSYHOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzene-1,3-dithiol vapor annealing was used for the improvement of power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Wu et al.
Nano letters, 10(5), 1628-1631 (2010-04-10)
Improvement in power conversion efficiency has been observed in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells through benzene-1,3-dithiol chemical vapor annealing. Phosphor NMR studies of the nanorods and TEM/AFM characterizations of the morphology of the blended film showed that the ligand
Aukse Navaruckiene et al.
Materials (Basel, Switzerland), 14(3) (2021-02-13)
The investigation of the influence of vanillin acrylate-based resin composition on photocuring kinetics and antimicrobial properties of the resulting polymers was performed in order to find efficient photocurable systems for optical 3D printing of bio-based polymers with tunable rigidity, as
Xinyu Tian et al.
ChemPlusChem, 85(8), 1639-1645 (2020-08-05)
We report the synthesis, characterization, and photophysical and ion-binding properties of deep-red to near-infrared (NIR) fluorescent rhodamine derivatives, bearing two spirolactone rings and substitution of the oxygen atoms in the xanthene ring with sulfur atoms (1-S). The diastereoisomeric cis- and
Atanu Ghosh et al.
Nanoscale, 9(3), 1068-1077 (2016-12-03)
We report the synthesis and unique reactivity of a new green dithiol protected cluster (DTPC), Ag
Dong-Kyun Ko et al.
ACS nano, 10(3), 3382-3388 (2016-02-26)
Recent advances in quantum dot surface passivation have led to a rapid development of high-efficiency solar cells. Another critical element for achieving efficient power conversion is the charge neutrality of quantum dots, as charge imbalances induce electronic states inside the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service