Skip to Content
Merck
All Photos(1)

Key Documents

203572

Sigma-Aldrich

Lead(II) chloride

99.999% trace metals basis

Synonym(s):

Cotunnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCl2
CAS Number:
Molecular Weight:
278.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks
solubility:
aliphatic hydrocarbons: slightly soluble(lit.)

grade

for analytical purposes

vapor pressure

1 mmHg ( 547 °C)

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

impurities

≤15.0 ppm Trace Metal Analysis

bp

950 °C (lit.)

mp

501 °C (lit.)

solubility

aliphatic hydrocarbons: slightly soluble(lit.)

density

5.85 g/mL at 25 °C (lit.)

SMILES string

Cl[PbH2]Cl

InChI

1S/2ClH.Pb/h2*1H;/q;;+2/p-2

InChI key

HWSZZLVAJGOAAY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Prepared by reacting lead(II) oxide /acetate or carbonate with HCl. In crystalline PbCl2, each atom is coordinated by nine Cl atoms, six of which lie at the apices of a trigonal prism and the remaining three beyond the centers of the three prism faces. Each Cl is coordinated by four or five Pb atoms. Upon exposure to air it form basic chlorides such as PbCl2.Pb(OH)2.

Application

Lead(II) chloride (PbCl2) (99.999%) is a lead halide that can be used in the formation of a precursor solution to form perovskite films. These films and sheets can be used in the fabrication of hybride solar absorbers and photodetectors. It also has potential application as a thermoelectric material.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Kawser Ahmed et al.
Environmental monitoring and assessment, 182(1-4), 197-204 (2011-01-07)
The present investigation was undertaken to study the induction of DNA damage by lead chloride (PbCl(2)) in freshwater climbing perch Anabas testudineus using alkaline single cell gel electrophoresis (comet assay). Based on the LC(50) values of lead chloride of A.
Travis Rush et al.
Neurotoxicology, 30(1), 47-51 (2008-11-26)
Chelation therapy for the treatment of acute, high dose exposure to heavy metals is accepted medical practice. However, a much wider use of metal chelators is by alternative health practitioners for so called "chelation therapy". Given this widespread and largely
Issa Tapsoba et al.
Analytical chemistry, 82(2), 457-460 (2009-12-25)
Lead-based compounds were used during antiquity as both pigments and medicines in the formulation of makeup materials. Chemical analysis of cosmetics samples found in Egyptians tombs and the reconstitution of ancient recipes as reported by Greco-Roman authors have shown that
Tiziana Gugliotta et al.
Cellular & molecular biology letters, 17(4), 586-597 (2012-09-04)
Our study concerns the effects of exposure to lead chloride on the morphology, K(+) efflux, SO(4)(-) influx and GSH levels of the human erythrocyte. Blood was collected in heparinized tubes and washed three times. The cells were suspended at 3%
Rustam Sadykov et al.
Journal of occupational health, 51(1), 64-73 (2008-12-20)
Lead's (Pb(II)) possible role in intestinal pathologies of microbial etiology remains mostly unknown. The aim of this study was to examine the effects of lead on the gut microbial community and its interactions with rat intestinal epithelium. The lead-induced changes

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service