Skip to Content
Merck
All Photos(1)

Key Documents

263826

Sigma-Aldrich

4-Iodobenzaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CHO
CAS Number:
Molecular Weight:
232.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

78-82 °C (lit.)

functional group

aldehyde
iodo

storage temp.

2-8°C

SMILES string

Ic1ccc(C=O)cc1

InChI

1S/C7H5IO/c8-7-3-1-6(5-9)2-4-7/h1-5H

InChI key

NIEBHDXUIJSHSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Iodobenzaldehyde participates in Suzuki–Miyaura coupling reaction to produce Metal–Organic Framework catalyst.

Application

4-Iodobenzaldehyde has been used in the preparation of:
  • benzaldimine monolayers
  • 4-[2-(trimethylsilyl)ethynyl]benzaldehyde
  • 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin
  • 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N′-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin, multipigment building block

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trans-Substituted porphyrin building blocks bearing iodo and ethynyl groups for applications in bioorganic and materials chemistry.
Ravikanth M, et al.
Tetrahedron, 54(27), 7721-7734 (1998)
Amino-Induced 2D Cu-Based Metal--Organic Framework as an Efficient Heterogeneous Catalyst for Aerobic Oxidation of Olefins
Tang J, et al.
Chemistry?A European Journal , 26, 4333-4340 (2020)
Umar Ali Dar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 240, 118570-118570 (2020-06-27)
The current study determines optical and fluorescence response of halogen substituted series of meso‑tetrakis‑(4‑halophenyl) porphyrin; H2TXPP (Halo = F, Cl, Br, I) dye in tetrahydrofuran; THF and THF-water system at changing pH in relationship with changing medium of allure. Effects produced
Selective cleavage of the carbon-halide bond in substituted benzaldimine monolayers by synchrotron soft X-ray: Anomalously large cleavage rate of the carbon-bromide bond.
Moon JH, et al.
Langmuir, 16(6), 2981-2984 (2000)
Abel T Demissie et al.
Journal of the American Chemical Society, 137(27), 8819-8828 (2015-06-23)
We report the systematic characterization of anisotropic, π-conjugated oligophenyleneimine (OPI) films synthesized using stepwise imine condensation, or "click" chemistry. Film synthesis began with a self-assembled monolayer (SAM) of 4-formylthiophenol or 4-aminothiophenol on Au, followed by repetitive, alternate addition of terephthalaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service