Skip to Content
Merck
All Photos(1)

Key Documents

255688

Sigma-Aldrich

1-Amino-4-methylpiperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13N3
CAS Number:
Molecular Weight:
115.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

172-175 °C (lit.)

solubility

water: soluble

density

0.957 g/mL at 25 °C (lit.)

SMILES string

CN1CCN(N)CC1

InChI

1S/C5H13N3/c1-7-2-4-8(6)5-3-7/h2-6H2,1H3

InChI key

RJWLLQWLBMJCFD-UHFFFAOYSA-N

Application

1-Amino-4-methylpiperazine has been used:
  • in synthesis of pyrazolo-pyrazine, -pyridine derivatives and 2-[(4-methylpiperazin-1-yl)iminomethyl]phenol
  • as a solvating and stabilizing agent in the preparation of uniform Ag nanoparticles (<10nm)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and stabilization of Ag nanoparticles in 1-amino-4-methylpiperazine.
Park H-S, et al.
Bull. Korean Chem. Soc., 32(1), 273-273 (2011)
2-[(4-Methylpiperazin-1-yl) iminomethyl] phenol.
Guo, M-L.
Acta Crystallographica Section E, Structure Reports Online, 63(4), o1788-o1789 (2007)
Henryk Foks et al.
Farmaco (Societa chimica italiana : 1989), 60(6-7), 513-517 (2005-06-14)
The investigations of new pyrazine and pyridine derivatives showing an antibacterial activity have been made. Upon treatment of 3-chloro-2-cyanopyrazine [1] and 2-chloro-3-cyanopyridine with 1,1-dimethyl-hydrazine, 1-aminopiperidine and 1-amino-4-methylpiperazine, either the pyrazolo-pyrazine (1), and -pyridine (2) derivatives, or ammonium salts (3-8) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service