Skip to Content
Merck
All Photos(2)

Key Documents

D179302

Sigma-Aldrich

1,4-Dimethylpiperazine

98%

Synonym(s):

1,4-Dimethylpiperazine, Lupetazine, N,N′-Dimethylpiperazine, Texacat DMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4463 (lit.)

bp

131-132 °C/750 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CN1CCN(C)CC1

InChI

1S/C6H14N2/c1-7-3-5-8(2)6-4-7/h3-6H2,1-2H3

InChI key

RXYPXQSKLGGKOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for polyurethane foams and intermediate for cationic surfactants.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beth Sharkey et al.
mAbs, 9(2), 257-268 (2016-12-13)
Monovalent bispecific antibodies (BsAbs) are projected to have broad clinical applications due to their ability to bind two different targets simultaneously. Although they can be produced using recombinant technologies, the correct pairing of heavy and light chains is a significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service