Skip to Content
Merck
All Photos(3)

Documents

208302

Sigma-Aldrich

Lead(II) thiocyanate

99.5% trace metals basis

Synonym(s):

Lead sulfocyanide, Lead(II) rhodanide, Plumbous thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(SCN)2
CAS Number:
Molecular Weight:
323.36
Beilstein:
3687984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99.5% trace metals basis

form

powder

mp

190 °C (dec.) (lit.)

solubility

cold water: soluble 200 part(lit.)
water: soluble 50 part (boiling)(lit.)

density

3.82 g/mL at 25 °C (lit.)

SMILES string

N#CS[PbH2]SC#N

InChI

1S/2CHNS.Pb/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

VRNINGUKUJWZTH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead (II) thiocyanate can be used:
  • To form a complex with Schiff-base lariat crown ether, N,N′-bis(3-(salicylaldimino)benzyl)-4,13-diaza-18-crown-6.
  • To investigate complexing and fluorescence properties of calixarene bearing two dansyl fluorophores grafted on a large pore mesoporous silica material.
  • As a reagent in the 1,2-dithiocyanation of alkynes in the presence of (dichloroiodo)benzene.
  • As a precursor to synthesize perovskite films for solar cell applications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mesoporous silica functionalized by a covalently bound calixarene-based fluoroionophore for selective optical sensing of mercury (II) in water
M'etivier R, et al.
Journal of Materials Chemistry, 15(27-28), 2965-2973 (2005)
(Dichloroiodo) benzene and lead (II) thiocyanate as an efficient reagent combination for stereoselective 1, 2-dithiocyanation of alkynes
Prakash O, et al.
Tetrahedron Letters, 42(4), 553-555 (2001)
A mesoporous silica functionalized by a covalently bound calixarene-based fluoroionophore for selective optical sensing of mercury (II) in water.
Metivier R, et al.
Journal of Materials Chemistry, 15(27-28), 2695-2673 (2005)
Carlos Platas-Iglesias et al.
Inorganic chemistry, 44(7), 2224-2233 (2005-03-29)
Compounds of formula [Pb(L2)(NCS)2] (1) and [Pb(L4)(SCN)2] (2) (where L2 is the lariat crown ether N,N'-bis(3-aminobenzyl)-4,13-diaza-18-crown-6 and L4 is the Schiff-base lariat crown ether N,N'-bis(3-(salicylaldimino)benzyl)-4,13-diaza-18-crown-6) were isolated and structurally characterized by X-ray diffraction analyses. The X-ray crystal structures of both
O Prakash et al.
The Journal of organic chemistry, 66(6), 2019-2023 (2001-04-13)
The combination reagent (dichloroiodo)benzene and lead(II) thiocyanate in dichloromethane effects oxidation of various enol silyl ethers, ketene silyl acetals, and beta-dicarbonyl compounds, thereby providing an efficient and convenient method for alpha-thiocyanation of carbonyl and beta-dicarbonyl compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service