251437
Rubidium carbonate
99.8% trace metals basis
Synonym(s):
Carbonic acid dirubidium, Dirubidium carboxide
About This Item
Recommended Products
grade
for analytical purposes
Assay
99.8% trace metals basis
form
powder
impurities
≤2500.0 ppm Trace Metal Analysis
SMILES string
[Rb+].[Rb+].[O-]C([O-])=O
InChI
1S/CH2O3.2Rb/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
InChI key
WPFGFHJALYCVMO-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1C - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service