Skip to Content
Merck
All Photos(3)

Documents

248924

Sigma-Aldrich

7-Amino-4-(trifluoromethyl)coumarin

≥99%

Synonym(s):

Coumarin 151

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F3NO2
CAS Number:
Molecular Weight:
229.16
Beilstein:
4456797
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥99%

form

(Powder or Crystals or Chunks)

mp

221-222 °C (lit.)

λmax

≤207 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C10H6F3NO2/c11-10(12,13)7-4-9(15)16-8-3-5(14)1-2-6(7)8/h1-4H,14H2

InChI key

JBNOVHJXQSHGRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Amino-4-(trifluoromethyl)coumarin (AFC) is a novel fluorescent marker used for the detection of proteinases.

Application

Suitable as laser dye

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and chemistry of 7-amino-4-(trifluoromethyl)coumarin and its amino acid and peptide derivatives
Bissell E R
The Journal of Organic Chemistry, 45, 2283?2287-2283?2287 (1980)
E D Pederson et al.
Journal of clinical periodontology, 21(8), 519-525 (1994-09-01)
Treponema denticola (Td) and Porphyromonas gingivalis (Pg) are associated with human moderate and severe adult periodontal diseases. This study quantifies these two anaerobes and their trypsin-like (TL) activities in subgingival plaque collected from both clinically healthy and periodontally diseased sites
S W Cox et al.
Journal of periodontal research, 24(6), 353-361 (1989-11-01)
Crevicular fluid samples were collected from 20 gingivitis and periodontitis patients using filter paper strips; these were then eluted into buffer. Portions of each sample were combined and the activities of this pooled eluate against different peptidyl derivatives of 7-amino-4-trifluoromethyl
Heike Helmbach et al.
The Journal of investigative dermatology, 118(6), 923-932 (2002-06-13)
Anticancer drugs kill susceptible cells through induction of apoptosis. Alterations of apoptotic pathways in drug-resistant tumor cells leading to apoptosis deficiency might represent a potent mechanism conferring drug resistance. We have assessed the effect of etoposide and cisplatin on the
Zhimin Tao et al.
Molecular pharmaceutics, 4(4), 583-595 (2007-04-19)
This study measures the time-dependence of cellular caspase activation by anticancer drugs and compares it with that of cellular respiration. Intracellular caspase activation and cellular respiration were measured during continuous exposure of Jurkat, HL-60, and HL-60/MX2 (deficient in topoisomerase-II) cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service