Skip to Content
Merck
All Photos(1)

Key Documents

247871

Sigma-Aldrich

2-Hydroxy-5-nitrobenzoic acid

99%

Synonym(s):

5-Nitrosalicylic acid, Anilotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
183.12
Beilstein:
2213719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

228-230 °C (lit.)

solubility

water: soluble 1g in 1475ml(lit.)
alcohol: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
hot water: soluble(lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1cc(ccc1O)[N+]([O-])=O

InChI

1S/C7H5NO5/c9-6-2-1-4(8(12)13)3-5(6)7(10)11/h1-3,9H,(H,10,11)

InChI key

PPDRLQLKHRZIJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxy-5-nitrobenzoic acid inhibits the activity of wild type-bovine low Mr protein tyrosine phosphatase. The inhibition constant has been evaluated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zhou et al.
Biochemistry, 38(9), 2636-2646 (1999-03-03)
Pyridoxal 5'-phosphate (PLP) binds tightly to bovine low Mr protein tyrosine phosphatase (BPTP), but it is a very poor substrate for the enzyme. The structural basis of this tight binding of PLP is examined here by a variety of methods.
Keishiro Nagoshi et al.
Journal of the American Society for Mass Spectrometry, 29(7), 1473-1483 (2018-04-21)
Radical-driven dissociation (RDD) of hydrogen-deficient peptide ions [M - H + H]·+ has been examined using matrix-assisted laser dissociation/ionization in-source decay mass spectrometry (MALDI-ISD MS) with the hydrogen-abstracting matrices 4-nitro-1-naphthol (4,1-NNL) and 5-nitrosalicylic acid (5-NSA). The preferential fragment ions observed in the ISD spectra
Sookjin Kang et al.
Free radical biology & medicine, 108, 715-724 (2017-05-06)
Mesalazine (5-aminosalicylic acid, 5-ASA), a currently used drug for anti-inflammatory bowel disease, is easily oxidized by HOCl, a strong oxidant generated in gut inflammation, to produce electrophilic quinones. We investigated whether this chemical feature has an implication in the anti-inflammatory
David Calligaris et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(32), 9978-9983 (2015-07-29)
We present a proof of concept study designed to support the clinical development of mass spectrometry imaging (MSI) for the detection of pituitary tumors during surgery. We analyzed by matrix-assisted laser desorption/ionization (MALDI) MSI six nonpathological (NP) human pituitary glands
Timotej Strmeň et al.
Rapid communications in mass spectrometry : RCM, 34(10), e8722-e8722 (2020-01-09)
Hyphenation of atmospheric pressure chemical ionization (APCI) mass spectrometry with capillary and micro high-performance liquid chromatography (HPLC) is attractive for many applications, but reliable ion sources dedicated to these conditions are still missing. There are a number of aspects to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service